Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460630_m1
          See other CHRNA2 GE Assays ›
          Gene Symbol:
          Chrna2
          Gene Name:
          cholinergic receptor, nicotinic, alpha polypeptide 2 (neuronal)
          Gene Aliases:
          Acra-2, Acra2, BC011490
          Chromosome Location:
          Chr.14: 66133238 - 66153396 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460630_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrna2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144803.2 NP_659052.1 6 - 7 1525 63
          GenBank mRNA AY574261.1 - 5 - 6 1409 63
          BC011490.1 - 6 - 7 1525 4193582 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000944.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrna2

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          cation transport
          signal transduction
          chemical synaptic transmission
          protein heterooligomerization
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          ion channel activity
          extracellular ligand-gated ion channel activity
          drug binding
          acetylcholine receptor activity
          acetylcholine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177513 BC011490.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2177513 NM_144803.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893229 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 919569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 699021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 878463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 287032 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86333 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168603 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124284 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177513 BC011490.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2177513 NM_144803.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398021 BC011490.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P398021 NM_144803.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29677633 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168603 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08144306 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124284 - -
          Whole Mouse Genome G4122A A_51_P398021 BC011490.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P398021 NM_144803.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124452 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168603 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124284 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11225567 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11180908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11225569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11200184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11200185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29677622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11407549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677636 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11347853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11459218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11333906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11627069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11200188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677644 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11627065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11627067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11200184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677648 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677622 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11347847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677632 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677638 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11751187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11347849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11225569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29677635 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10416080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416077 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10416079 - -
          Affymetrix HT MG-430 PM Array Plate 1425409_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10416071 - -
          Mouse Genome 430 2.0 Array 1425409_PM_at - -
          Mouse Genome 430 2.0 Array 1425409_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110902_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110902_at - -
          Affymetrix Mouse Clariom S Array PSR1400007585.mm.1 NM_144803.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400007585.mm.1 NM_144803.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400007585.mm.1 TC1400000944.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1425409_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425409_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 426110 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050315 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050315 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2681916 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2681916 - -
          mouse-6 v1.1 (Illumina) scl46158.7.1_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050315 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46158.7.1_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050315 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2681916 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2681916 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline