Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460745_m1
          See other FRS3 GE Assays ›
          Gene Symbol:
          Frs3
          Gene Name:
          fibroblast growth factor receptor substrate 3
          Gene Aliases:
          4930417B13Rik, AI449674, Frs2beta, Snt2
          Chromosome Location:
          Chr.17: 47687296 - 47704286 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460745_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Frs3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144939.2 NP_659188.1 5 - 6 648 105
          GenBank mRNA BC014819.1 - 5 - 6 648 4506982 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002793.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Frs3

          Gene Ontology Categories:

          Function(s) Process(es)

          fibroblast growth factor receptor signaling pathway
          fibroblast growth factor receptor binding
          insulin receptor binding
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31213685 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31213686 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020736 BC014819.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020736 NM_144939.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2020736 AK077209.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 92514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872533 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48497 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111153 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020736 BC014819.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020736 NM_144939.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2020736 AK077209.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P444162 BC014819.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31213695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03326948 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111153 - -
          Whole Mouse Genome G4122A A_51_P444162 BC014819.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126633 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111153 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13432307 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31213685 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31213686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31213820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13218885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213699 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213805 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13144724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13103197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13204189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13410852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13414044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213751 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13432307 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13096605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13203869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213801 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13414055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13133296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213691 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13218884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13096608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13204194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13414070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13575498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13250909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13148159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213680 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13346340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13144717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13096604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13414059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13196769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213798 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13557378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31213731 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445716 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445717 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445715 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445714 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445713 - -
          Affymetrix HT MG-430 PM Array Plate 1424449_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445710 - -
          Mouse Genome 430 2.0 Array 1439277_at - -
          Mouse Genome 430 2.0 Array 1424449_PM_at - -
          Mouse Genome 430 2.0 Array 1439277_PM_at - -
          Mouse Genome 430 2.0 Array 1424449_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107971_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107971_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700008897.mm.1 NM_144939.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008897.mm.1 NM_144939.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700008897.mm.1 TC1700002793.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1424449_at - -
          Affymetrix HT Mouse Genome 430A Array 1424449_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477069 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5890048 - -
          Illumina mouse-6 v1.0 expression beadchip 5890048 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005890048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2638186 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2638186 - -
          mouse-6 v1.1 (Illumina) scl50645.13_521-S - -
          Illumina Sentrix mouse-6.v1 bead array 5890048 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50645.13_521-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5890048 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2638186 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2638186 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50645.13_521-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline