Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00460886_m1
          See other GTPBP6 GE Assays ›
          Gene Symbol:
          Gtpbp6
          Gene Name:
          GTP binding protein 6 (putative)
          Gene Aliases:
          AV119224, Pgbpll, pgpl
          Chromosome Location:
          Chr.5: 110103976 - 110108197 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00460886_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gtpbp6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_145147.5 NP_660129.2 5 - 6 745 136
          GenBank mRNA AJ293619.1 - 4 - 5 375 136
          AK082192.1 - 2 - 3 621 136
          AK144812.1 - 5 - 6 746 136
          AK152225.1 - 5 - 6 746 136
          AK152972.1 - 5 - 6 746 136
          AK159462.1 - 5 - 6 718 136
          BC027821.1 - 2 - 3 276 5321725 136

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002981.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gtpbp6

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          molecular_function
          GTP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977816 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977818 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977819 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977813 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968483 BC034116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968483 NR_027665.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968483 NM_207279.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968483 AK152225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968483 AK152972.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973801 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167937 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968483 BC034116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968483 NR_027665.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968483 NM_207279.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968483 AK152225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968483 AK152972.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23977807 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167937 - -
          Whole Mouse Genome G4122A A_52_P380250 AK149435.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380250 BC034116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380250 NR_027665.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380250 NM_207279.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P380250 AK152225.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133754 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167937 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890562 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04890566 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977819 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977813 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977824 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977816 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977825 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977814 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977818 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977823 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977815 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23977822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23977853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04543212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04150703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04149933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04183194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04177267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04848684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04890583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977853 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04177261 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23977809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532321 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532328 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532326 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532323 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532322 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532324 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532319 - -
          Affymetrix HT MG-430 PM Array Plate 1427350_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532317 - -
          Mouse Genome 430 2.0 Array 1427350_a_at - -
          Mouse Genome 430 2.0 Array 1427350_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107999_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107999_at - -
          Affymetrix Mouse Clariom S Array PSR0500026344.mm.1 TC0500002981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026344.mm.1 NM_145147.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026344.mm.1 NM_145147.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026345.mm.1 TC0500002981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026345.mm.1 NM_145147.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026345.mm.1 NM_145147.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026346.mm.1 TC0500002981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026346.mm.1 NM_145147.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026346.mm.1 NM_145147.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026349.mm.1 TC0500002981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026349.mm.1 NM_145147.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026349.mm.1 NM_145147.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026352.mm.1 TC0500002981.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026352.mm.1 NM_145147.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026352.mm.1 NM_145147.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026355.mm.1 TC0500002981.mm.1 MTA TC
          Murine Genome U74v2 Set 171161_PM_i_at - -
          Murine Genome U74v2 Set 171161_i_at - -
          Affymetrix HT Mouse Genome 430A Array 1427350_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427350_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 670471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000670471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213272 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213272 - -
          mouse-6 v1.1 (Illumina) scl26235.9.1_65-S - -
          Illumina Sentrix mouse-6.v1 bead array 670471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26235.9.1_65-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2901056 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213272 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2901056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl26235.9.1_65-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline