Search
Search
| Gene Symbol: Oas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145226.2 | NP_660261.1 | 12 - 13 | 2879 | 86 | |
| GenBank mRNA | AB067534.1 | - | 12 - 13 | 2879 | 86 | |
| AF453830.1 | - | 12 - 13 | 2871 | 86 | ||
| BC016461.1 | - | 5 - 6 | 977 | 86 | ||
| BC141055.1 | - | 12 - 13 | 2872 | 9055667 | 86 | |
Set Membership: |
|