Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461447_g1
          See other ZFP768 GE Assays ›
          Gene Symbol:
          Zfp768
          Gene Name:
          zinc finger protein 768
          Gene Aliases:
          BC026432, Znf768
          Chromosome Location:
          Chr.7: 127342795 - 127345314 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461447_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp768
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146202.1 NP_666314.1 1 - 2 168 57
          GenBank mRNA AK155155.1 - 1 - 2 280 57
          BC026432.1 - 1 - 2 168 4235387 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004323.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfp768

          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          poly(A) RNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505277 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505270 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505276 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505278 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 AK080841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 AK155155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 BC026432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 NM_146202.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 AK080841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 AK155155.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 BC026432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P256224 NM_146202.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845324 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 719065 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 56562 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P173789 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 AK080841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 AK155155.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 BC026432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 NM_146202.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 AK080841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 AK155155.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 BC026432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P256224 NM_146202.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25505259 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P173789 - -
          Whole Mouse Genome G4122A A_51_P256224 AK080841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 AK155155.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 BC026432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 NM_146202.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 AK080841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 AK155155.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 BC026432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P256224 NM_146202.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139782 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P173789 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200648 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200661 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06200623 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505274 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505277 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505269 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505271 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505266 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505268 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505270 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505248 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505251 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505273 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25505276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25505270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06231068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05896325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05896318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05896324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06151824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06470049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06230218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06170396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06200641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505304 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505309 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568227 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568226 - -
          Affymetrix HT MG-430 PM Array Plate 1424506_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568225 - -
          Mouse Genome 430 2.0 Array 1424506_PM_at - -
          Mouse Genome 430 2.0 Array 1424506_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233890_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233890_at - -
          Affymetrix Mouse Clariom S Array PSR0700036302.mm.1 NM_146202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036302.mm.1 NM_146202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036302.mm.1 TC0700004323.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036301.mm.1 NM_146202.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036301.mm.1 NM_146202.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036301.mm.1 TC0700004323.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1424506_at - -
          Affymetrix HT Mouse Genome 430A Array 1424506_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 616293 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 670239 XM_001479063.1 refseq_mrna_mouse_XM
          Illumina mouse-6 v1.0 expression beadchip 670239 XM_001479063.1 refseq_mrna_mouse_XM
          Illumina mouseRef-8 v1.0 expression beadchip 0000670239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2721919 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2721919 - -
          mouse-6 v1.1 (Illumina) scl30650.2_250-S - -
          Illumina Sentrix mouse-6.v1 bead array 670239 XM_001479063.1 refseq_mrna_mouse_XM
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30650.2_250-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 670239 XM_001479063.1 refseq_mrna_mouse_XM
          MouseRef-8 v2 Expression BeadChip ILMN_2721919 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2721919 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline