Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461575_m1
          See other TBC1D13 GE Assays ›
          Gene Symbol:
          Tbc1d13
          Gene Name:
          TBC1 domain family, member 13
          Gene Aliases:
          2600014A06Rik, BC025586
          Chromosome Location:
          Chr.2: 30133751 - 30152013 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461575_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tbc1d13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146252.2 NP_666364.1 6 - 7 399 81
          GenBank mRNA AK158066.1 - 6 - 7 480 81
          BC025586.1 - 6 - 7 425 5324875 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000525.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tbc1d13

          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          GTPase activator activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101537 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101536 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102047 BC025586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102047 BC039215.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102047 AK158066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102047 NM_146252.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 561581 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 868327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 877041 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 162876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135918 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146071 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137538 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102047 BC025586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102047 BC039215.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102047 AK158066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P102047 NM_146252.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101550 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101554 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101543 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101552 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146071 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137538 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04470170 - -
          Whole Mouse Genome G4122A A_51_P218165 BC025586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P218165 NM_146252.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102047 BC025586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102047 BC039215.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102047 AK158066.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102047 NM_146252.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146071 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137538 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01163678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01163672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101536 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101538 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02168927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01824594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101574 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101560 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101527 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101534 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101538 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01163686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01991440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01966601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02168921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01991443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01643847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01163679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101536 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470938 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470937 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470942 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470943 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470940 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470939 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470941 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470944 - -
          Affymetrix HT MG-430 PM Array Plate 1424082_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470936 - -
          Mouse Genome 430 2.0 Array 1442717_PM_at - -
          Mouse Genome 430 2.0 Array 1424082_at - -
          Mouse Genome 430 2.0 Array 1442717_at - -
          Mouse Genome 430 2.0 Array 1424082_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70296_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70296_at - -
          Affymetrix Mouse Clariom S Array PSR0200004108.mm.1 NM_146252.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200004108.mm.1 TC0200000525.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1424082_at - -
          Affymetrix HT Mouse Genome 430A Array 1424082_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 317885 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 870010 - -
          Illumina mouse-6 v1.0 expression beadchip 870010 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000870010 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218901 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218901 - -
          mouse-6 v1.1 (Illumina) scl21096.14_260-S - -
          Illumina Sentrix mouse-6.v1 bead array 870010 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21096.14_260-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 870010 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218901 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218901 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21096.14_260-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-knt4q:80/100.66.75.14:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0