Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461740_g1
          See other LY6G5B GE Assays ›
          Gene Symbol:
          Ly6g5b
          Gene Name:
          lymphocyte antigen 6 complex, locus G5B
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 35113944 - 35115400 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461740_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ly6g5b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_148939.2 NP_683741.1 1 - 2 58 65
          GenBank mRNA AJ315553.1 - 1 - 2 58 65
          BC131964.1 - 1 - 2 86 40130241 65
          BC131966.1 - 1 - 2 86 40130243 65
          HE864438.1 - 1 - 2 74 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001946.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ly6g5b

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972133 AK151902.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972133 BC131964.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972133 BC131966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972133 BC145150.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972133 NM_148939.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245765 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146921 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972133 AK151902.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972133 BC131964.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972133 BC131966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972133 BC145150.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972133 NM_148939.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369443 AK151902.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369443 BC131964.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369443 BC131966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369443 BC145150.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369443 NM_148939.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31157668 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146921 - -
          Whole Mouse Genome G4122A A_51_P369443 AK151902.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369443 BC131964.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369443 BC131966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369443 BC145150.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369443 NM_148939.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111885 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146921 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13505575 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13505576 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13505595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13505593 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13505578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31157675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13090435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13505582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157659 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157656 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13505595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157652 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31157657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13505573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13505600 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450433 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450431 - -
          Affymetrix Mouse Clariom S Array PSR1700018474.mm.1 NM_148939.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018474.mm.1 NM_148939.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018474.mm.1 TC1700001946.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018476.mm.1 NM_148939.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018476.mm.1 NM_148939.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700018476.mm.1 TC1700001946.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700018477.mm.1 TC1700001946.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352609 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6110129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2608000 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2608000 - -
          mouse-6 v1.1 (Illumina) scl49997.2.1_244-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49997.2.1_244-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2608000 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2608000 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline