Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461745_m1
          See other GIPC3 GE Assays ›
          Gene Symbol:
          Gipc3
          Gene Name:
          GIPC PDZ domain containing family, member 3
          Gene Aliases:
          Ahl5, Rgs19ip3
          Chromosome Location:
          Chr.10: 81336540 - 81343744 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461745_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gipc3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_148951.1 NP_683753.1 4 - 5 668 80
          GenBank mRNA AB074494.1 - 4 - 5 668 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002537.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gipc3

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s)

          biological_process

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289009 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288984 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289007 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289008 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079991 AB074494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2079991 NM_148951.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 880944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142840 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140786 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079991 AB074494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2079991 NM_148951.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P102005 AB074494.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P102005 NM_148951.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289002 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289003 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27288999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289006 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27289008 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07125317 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140786 - -
          Whole Mouse Genome G4122A A_51_P102005 AB074494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102005 NM_148951.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105548 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140786 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08832063 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08429503 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289006 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289007 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289009 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288986 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27289008 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27288984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27289007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27288961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08604464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08849571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08849585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08849583 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08604462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08385021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08610344 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08224572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08832063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27288971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27289006 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10371139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10371136 - -
          Affymetrix HT MG-430 PM Array Plate 1426150_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10371134 - -
          Mouse Genome 430 2.0 Array 1426150_PM_at - -
          Mouse Genome 430 2.0 Array 1426150_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209047_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 209047_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000018991.mm.1 NM_148951.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018991.mm.1 NM_148951.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018991.mm.1 TC1000002537.mm.1 MTA TC
          Mouse Expression Set 430 1426150_PM_at - -
          Mouse Expression Set 430 1426150_at - -
          Mouse Genome 430A 2.0 Array 1426150_PM_at - -
          Mouse Genome 430A 2.0 Array 1426150_at - -
          Affymetrix HT Mouse Genome 430A Array 1426150_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426150_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 839928 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 770278 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000770278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770482 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770482 - -
          mouse-6 v1.1 (Illumina) scl0209047.3_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 770278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0209047.3_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 770278 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3001202 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770482 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3001202 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline