| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100010132.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010132.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010132.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010140.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010140.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010140.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010141.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010141.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010141.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010154.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010154.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010154.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010135.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010135.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010135.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010146.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010146.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010146.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010155.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010155.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010155.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010157.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010157.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010157.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010142.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010142.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010142.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010143.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010143.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010143.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010150.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010150.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010150.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010152.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010152.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010152.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010133.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010133.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010133.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010147.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010147.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010147.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010156.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010156.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010156.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010137.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010137.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010137.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010153.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010153.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010153.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010144.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010144.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010144.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010145.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010145.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010145.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010149.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010149.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010149.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010134.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010134.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010134.mm.1 |
TC0100001234.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100010151.mm.1 |
NM_152895.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010151.mm.1 |
NM_152895.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100010151.mm.1 |
TC0100001234.mm.1 |
MTA TC |