Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461807_g1
          See other EGFL8 GE Assays ›
          Gene Symbol:
          Egfl8
          Gene Name:
          EGF-like domain 8
          Gene Aliases:
          Ng3
          Chromosome Location:
          Chr.17: 34613200 - 34616572 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461807_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Egfl8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152922.3 NP_690886.3 2 - 3 191 78
          GenBank mRNA AK148723.1 - 2 - 3 192 78
          BC141312.1 - 2 - 3 160 9055924 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002812.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Egfl8

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          calcium ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 BC055829.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 NM_152922.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AK148723.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AY635583.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115507 AK086281.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200457 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142370 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 BC055829.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 NM_152922.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AK148723.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AY635583.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115507 AK086281.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31154827 - -
          Whole Mouse Genome G4122A A_51_P225112 BC141312.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225112 NM_152922.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P225112 AK148723.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124539 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351463 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351455 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351454 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351429 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13351450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31154788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31154825 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450223 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450220 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450215 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450213 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450214 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450221 - -
          Affymetrix HT MG-430 PM Array Plate 1447953_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450212 - -
          Affymetrix Mouse Gene 1.1 ST Array 10444420 - -
          Mouse Genome 430 2.0 Array 1447953_at - -
          Mouse Genome 430 2.0 Array 1447953_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81701_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 81701_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700017918.mm.1 NM_152922.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017918.mm.1 NM_152922.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017918.mm.1 TC1700002812.mm.1 MTA TC
          Murine Genome U74v2 Set 101345_at - -
          Murine Genome U74v2 Set 101345_PM_at - -
          Murine Genome U74v2 Set 167187_PM_at - -
          Murine Genome U74v2 Set 167187_at - -
          Affymetrix HT Mouse Genome 430A Array 1447953_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1447953_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 763034 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 360102 - -
          Illumina mouse-6 v1.0 expression beadchip 1170647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170647 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2638839 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593189 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2593189 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2638839 - -
          mouse-6 v1.1 (Illumina) scl0001741.1_77-S - -
          mouse-6 v1.1 (Illumina) scl50018.6.1_12-S - -
          Illumina Sentrix mouse-6.v1 bead array 360102 - -
          Illumina Sentrix mouse-6.v1 bead array 1170647 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001741.1_77-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50018.6.1_12-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170647 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2593188 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2593187 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593187 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593188 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline