Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00461814_m1
          See other MMP21 GE Assays ›
          Gene Symbol:
          Mmp21
          Gene Name:
          matrix metallopeptidase 21
          Gene Aliases:
          b2b2458Clo, b2b873Clo
          Chromosome Location:
          Chr.7: 133671568 - 133682438 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00461814_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mmp21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_152944.1 NP_694423.1 1 - 2 178 74
          GenBank mRNA AF507967.1 - 1 - 2 163 74
          AY124569.1 - 1 - 2 178 74
          BC106860.2 - 1 - 2 178 40047272 74
          BC106861.2 - 1 - 2 178 40047279 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004427.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mmp21

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          hematopoietic progenitor cell differentiation
          proteolysis
          metalloendopeptidase activity
          calcium ion binding
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170841 BC106860.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170841 BC106861.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170841 BC111522.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170841 NM_152944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2170841 AY124569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046985 NM_152944.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2046985 AY124569.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 285641 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 371166 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046985 NM_152944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2046985 AY124569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170841 BC106860.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170841 BC106861.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170841 BC111522.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170841 NM_152944.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2170841 AY124569.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P272866 BC106860.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P272866 BC106861.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P272866 BC111522.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P272866 NM_152944.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P272866 AF507967.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542013 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25541992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25542032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01642026 - -
          Whole Mouse Genome G4122A A_51_P272866 BC106860.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272866 BC106861.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272866 BC111522.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272866 NM_152944.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P272866 AF507967.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115739 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139345 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06529140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930474 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06529137 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05930475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25541888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25542015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06029565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06410717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06616126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06333661 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06279538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06410711 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05930481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06647891 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06647882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06029552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06112275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25541979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25542020 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568637 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568633 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568630 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568629 - -
          Affymetrix Mouse Clariom S Array PSR0700037187.mm.1 NM_152944.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037187.mm.1 NM_152944.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037187.mm.1 TC0700004427.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037189.mm.1 NM_152944.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037189.mm.1 NM_152944.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037189.mm.1 TC0700004427.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 802355 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4480673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004480673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223199 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223199 - -
          mouse-6 v1.1 (Illumina) scl30563.7.1_113-S - -
          Illumina Sentrix mouse-6.v1 bead array 4480673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30563.7.1_113-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4480673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1223199 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1223199 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline