Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462068_m1
          See other THTPA GE Assays ›
          Gene Symbol:
          Thtpa
          Gene Name:
          thiamine triphosphatase
          Gene Aliases:
          AW122966
          Chromosome Location:
          Chr.14: 55094784 - 55098995 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462068_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Thtpa
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153083.5 NP_694723.1 1 - 2 1227 87
          GenBank mRNA AF432864.2 - 1 - 2 1058 87
          AK132479.1 - 1 - 2 919 87
          BC025562.1 - 1 - 2 1211 5322971 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400000772.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Thtpa

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          thiamine metabolic process
          dephosphorylation
          thiamine diphosphate metabolic process
          magnesium ion binding
          hydrolase activity
          metal ion binding
          thiamin-triphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 BC025562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK085173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK132479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK178674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 NM_153083.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 BC025562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK085173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK132479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 AK178674.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P674357 NM_153083.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600320 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13931 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178114 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113208 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 BC025562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK085173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK132479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK178674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 NM_153083.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 BC025562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK085173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK132479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 AK178674.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P674357 NM_153083.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29616238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113208 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178114 - -
          Whole Mouse Genome G4122A A_52_P674357 BC025562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK085173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK132479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK178674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 NM_153083.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 BC025562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK085173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK132479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 AK178674.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674357 NM_153083.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109267 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178114 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113208 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11217092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29616239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11217088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11217103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11747469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440651 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11217092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29616260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11440678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11217086 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10415160 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415162 - -
          Affymetrix Mouse Gene 1.0 ST Array 10415161 - -
          Affymetrix HT MG-430 PM Array Plate 1433436_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1428003_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10415159 - -
          Mouse Genome 430 2.0 Array 1428003_s_at - -
          Mouse Genome 430 2.0 Array 1433436_s_at - -
          Mouse Genome 430 2.0 Array 1428003_PM_s_at - -
          Mouse Genome 430 2.0 Array 1433436_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105663_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 105663_at - -
          Affymetrix Mouse Clariom S Array PSR1400006051.mm.1 NM_153083.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006051.mm.1 NM_153083.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006051.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006048.mm.1 NM_153083.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006048.mm.1 NM_153083.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006048.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006050.mm.1 NM_153083.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006050.mm.1 NM_153083.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006050.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006049.mm.1 NM_153083.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006049.mm.1 NM_153083.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006049.mm.1 TC1400000772.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400006052.mm.1 NM_153083.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006052.mm.1 NM_153083.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1400006052.mm.1 TC1400000772.mm.1 MTA TC
          Mouse Expression Set 430 1433436_s_at - -
          Mouse Expression Set 430 1433436_PM_s_at - -
          Mouse Expression Set 430 1428003_PM_s_at - -
          Mouse Expression Set 430 1428003_s_at - -
          Mouse Genome 430A 2.0 Array 1433436_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1433436_s_at - -
          Mouse Genome 430A 2.0 Array 1428003_s_at - -
          Mouse Genome 430A 2.0 Array 1428003_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 587418 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104150139 - -
          Illumina mouse-6 v1.0 expression beadchip 104150139 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2683095 - -
          mouse-6 v1.1 (Illumina) scl0105663.2_267-S - -
          Illumina Sentrix mouse-6.v1 bead array 104150139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104150139 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2794825 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2794825 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2683095 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline