Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462071_g1
          See other TMEM191C GE Assays ›
          Gene Symbol:
          Tmem191c
          Gene Name:
          transmembrane protein 191C
          Gene Aliases:
          4933405M22Rik, D16Bwg1494e, MNCb-4137
          Chromosome Location:
          Chr.16: 17276300 - 17278661 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462071_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem191c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177473.3 NP_803424.2 5 - 6 480 68
          GenBank mRNA AB041601.1 - 3 - 4 671 68
          BC058760.1 - 1 - 2 575 68
          BC098238.1 - 1 - 2 107 68
          BU936413.1 - 5 - 6 463 68
          BY296351.1 - 5 - 6 335 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000213.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB041601.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem191c

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BC058760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BU936413.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BY296351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 NM_177473.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 AB041601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BC058760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BU936413.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 BY296351.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 NM_177473.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P1516 AB041601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 AK016675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 BC058760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 BC098238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 NM_177473.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 AB041601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 AK016675.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 BC058760.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 BC098238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 NM_177473.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P165082 AB041601.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 891330 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333900 AF094479.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333900 BC018307.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 333900 NM_010010.1 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115039 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 AK016675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 BC058760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 BC098238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 NM_177473.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 AB041601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 AK016675.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 BC058760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 BC098238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 NM_177473.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P165082 AB041601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BC058760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BU936413.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BY296351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 NM_177473.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 AB041601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BC058760.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BU936413.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 BY296351.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 NM_177473.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P1516 AB041601.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115039 - -
          Whole Mouse Genome G4122A A_51_P165082 AK016675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 BC058760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 BC098238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 NM_177473.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 AB041601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 AK016675.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 BC058760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 BC098238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 NM_177473.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P165082 AB041601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BC058760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BU936413.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BY296351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 NM_177473.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 AB041601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BC058760.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BU936413.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 BY296351.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 NM_177473.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P1516 AB041601.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115039 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433978 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433980 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433979 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433987 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433982 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433984 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433985 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433983 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433986 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433981 - -
          Affymetrix HT MG-430 PM Array Plate 1420191_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1449770_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1420192_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1452234_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433977 - -
          Mouse Genome 430 2.0 Array 1452234_s_at - -
          Mouse Genome 430 2.0 Array 1420191_s_at - -
          Mouse Genome 430 2.0 Array 1420192_PM_at - -
          Mouse Genome 430 2.0 Array 1420192_at - -
          Mouse Genome 430 2.0 Array 1449770_x_at - -
          Mouse Genome 430 2.0 Array 1452234_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449770_PM_x_at - -
          Mouse Genome 430 2.0 Array 1420191_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224019_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224019_at - -
          Affymetrix Mouse Clariom S Array PSR1600001723.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001723.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001723.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001726.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001726.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001726.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001730.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001730.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001730.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001746.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001746.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001746.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001727.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001727.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001727.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001739.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001739.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001739.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001738.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001738.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001738.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001752.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001752.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001752.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001753.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001753.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001753.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001749.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001749.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001749.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001750.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001750.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001750.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001733.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001733.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001733.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001751.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001751.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001751.mm.1 TC1600000213.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600001743.mm.1 NM_177473.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001743.mm.1 NM_177473.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1600001743.mm.1 TC1600000213.mm.1 MTA TC
          Mouse Expression Set 430 1420192_PM_at - -
          Mouse Expression Set 430 1452234_PM_s_at - -
          Mouse Expression Set 430 1420191_s_at - -
          Mouse Expression Set 430 1420191_PM_s_at - -
          Mouse Expression Set 430 1420192_at - -
          Mouse Expression Set 430 1449770_PM_x_at - -
          Mouse Expression Set 430 1452234_s_at - -
          Mouse Expression Set 430 1449770_x_at - -
          Murine Genome U74v2 Set 160964_at - -
          Murine Genome U74v2 Set 161794_PM_i_at - -
          Murine Genome U74v2 Set 161794_i_at - -
          Murine Genome U74v2 Set 160964_PM_at - -
          Murine Genome U74v2 Set 166846_r_at - -
          Murine Genome U74v2 Set 166846_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1452234_s_at - -
          Mouse Genome 430A 2.0 Array 1452234_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1449770_x_at - -
          Mouse Genome 430A 2.0 Array 1420191_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1420191_s_at - -
          Mouse Genome 430A 2.0 Array 1449770_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1420192_at - -
          Mouse Genome 430A 2.0 Array 1420192_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449770_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420192_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420192_at - -
          Affymetrix HT Mouse Genome 430A Array 1449770_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1420191_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452234_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1420191_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452234_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 464396 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2704318 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline