Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462088_m1
          See other GJD4 GE Assays ›
          Gene Symbol:
          Gjd4
          Gene Name:
          gap junction protein, delta 4
          Gene Aliases:
          9430022F06Rik, Cx39
          Chromosome Location:
          Chr.18: 9137290 - 9289808 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gjd4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153086.5 NP_694726.2 1 - 2 248 72
          GenBank mRNA AJ414562.1 - 1 - 2 57 72
          AK034663.1 - 1 - 2 249 72
          AK053016.1 - 1 - 2 200 72
          BC137984.1 - 1 - 2 225 8861003 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000961.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gjd4

          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s) Process(es)

          cell communication
          regulation of satellite cell activation involved in skeletal muscle regeneration
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125346 AK034663.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2125346 NM_153086.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838129 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29375 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153190 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117838 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149453 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125346 AK034663.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2125346 NM_153086.5 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P157185 AK034663.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P157185 AK053016.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P157185 NM_153086.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31484836 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153190 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149453 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117838 - -
          Whole Mouse Genome G4122A A_51_P157185 AK034663.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157185 AK053016.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P157185 NM_153086.5 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153190 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117838 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13623184 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14108188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13623187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31484838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13769669 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13650202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13769682 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13843386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484813 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13594852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484823 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13980450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484852 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14133406 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484817 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13980443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484770 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13769687 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14134784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484784 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31484804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14134786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13634216 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10457384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10457383 - -
          Affymetrix HT MG-430 PM Array Plate 1427601_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10457382 - -
          Mouse Genome 430 2.0 Array 1427601_at - -
          Mouse Genome 430 2.0 Array 1427601_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225152_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 225152_at - -
          Affymetrix Mouse Clariom S Array PSR1800007112.mm.1 NM_153086.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007112.mm.1 NM_153086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007112.mm.1 TC1800000961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800007113.mm.1 NM_153086.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007113.mm.1 NM_153086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007113.mm.1 TC1800000961.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800007111.mm.1 NM_153086.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007111.mm.1 NM_153086.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1800007111.mm.1 TC1800000961.mm.1 MTA TC
          Mouse Expression Set 430 1427601_at - -
          Mouse Expression Set 430 1427601_PM_at - -
          Mouse Genome 430A 2.0 Array 1427601_PM_at - -
          Mouse Genome 430A 2.0 Array 1427601_at - -
          Affymetrix HT Mouse Genome 430A Array 1427601_at - -
          Affymetrix HT Mouse Genome 430A Array 1427601_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 588499 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3130253 - -
          Illumina mouse-6 v1.0 expression beadchip 3130253 - -
          Illumina mouse-6 v1.0 expression beadchip 6860750 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003130253 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860750 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2770767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2770767 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243047 - -
          mouse-6 v1.1 (Illumina) scl00225152.2_231-S - -
          mouse-6 v1.1 (Illumina) scl0225152.1_57-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860750 - -
          Illumina Sentrix mouse-6.v1 bead array 3130253 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00225152.2_231-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0225152.1_57-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3130253 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860750 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2770767 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243047 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline