Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462169_m1
          See other RTP3 GE Assays ›
          Gene Symbol:
          Rtp3
          Gene Name:
          receptor transporter protein 3
          Gene Aliases:
          Tmem7
          Chromosome Location:
          Chr.9: 110984938 - 110990578 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462169_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153100.2 NP_694740.2 1 - 2 273 68
          GenBank mRNA AJ428064.1 - 1 - 2 273 68
          AY562227.1 - 1 - 2 171 68
          BC112903.1 - 1 - 2 273 4196964 68
          BC150678.1 - 1 - 2 217 9087589 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003122.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtp3

          Gene Ontology Categories:

          Process(es)

          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 AY562227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 BC112903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 BC150678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 NM_153100.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 AJ428064.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 AY562227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 BC112903.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 BC150678.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 NM_153100.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P134023 AJ428064.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 86095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811255 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161811 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 AY562227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 BC112903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 BC150678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 NM_153100.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 AJ428064.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 AY562227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 BC112903.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 BC150678.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 NM_153100.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P134023 AJ428064.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827022 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827011 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827048 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26827032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P161811 - -
          Whole Mouse Genome G4122A A_52_P134023 AY562227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 BC112903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 BC150678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 NM_153100.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 AJ428064.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 AY562227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 BC112903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 BC150678.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 NM_153100.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P134023 AJ428064.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121186 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106803 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P161811 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07699679 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07699688 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07699697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26826992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26827027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07699693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07699683 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07758842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07699688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07598403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07936493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07851542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07598405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07781432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08170989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07851541 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26826993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07874403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08019581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07598408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26827060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597277 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597276 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589696 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597275 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597274 - -
          Affymetrix Mouse Gene 1.0 ST Array 10589697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597278 - -
          Affymetrix HT MG-430 PM Array Plate 1452472_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10589695 - -
          Affymetrix Mouse Gene 1.1 ST Array 10597273 - -
          Mouse Genome 430 2.0 Array 1452472_PM_at - -
          Mouse Genome 430 2.0 Array 1452472_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235636_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235636_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900026446.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026446.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026446.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026447.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026447.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026447.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026456.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026456.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026456.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026459.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026459.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026459.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026449.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026449.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026449.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026458.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026458.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026458.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026453.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026457.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026457.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026457.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026460.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026460.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026460.mm.1 TC0900003122.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026445.mm.1 NM_153100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026445.mm.1 NM_153100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026445.mm.1 TC0900003122.mm.1 MTA TC
          Mouse Expression Set 430 1452472_at - -
          Mouse Expression Set 430 1452472_PM_at - -
          Mouse Genome 430A 2.0 Array 1452472_at - -
          Mouse Genome 430A 2.0 Array 1452472_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452472_at - -
          Affymetrix HT Mouse Genome 430A Array 1452472_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 720064 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2260176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1239067 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1239067 - -
          mouse-6 v1.1 (Illumina) scl35305.4.1_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35305.4.1_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2864416 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2864416 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1239067 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline