Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00462487_m1
          See other 2410015M20RIK GE Assays ›
          Gene Symbol:
          2410015M20Rik
          Gene Name:
          RIKEN cDNA 2410015M20 gene
          Gene Aliases:
          QIL1, sr104
          Chromosome Location:
          Chr.17: 56607452 - 56609771 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00462487_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2410015M20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153152.3 NP_694792.1 3 - 4 363 84
          GenBank mRNA AK075942.1 - 3 - 4 294 84
          AK075961.1 - 3 - 4 364 84
          AY091637.1 - 3 - 4 306 84
          BC049674.1 - 184 84
          BC056164.1 - 3 - 4 269 5713062 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002332.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 2410015M20Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257285 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 NM_153152.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 AK075942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 AK075961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 BC056164.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 NM_153152.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 AK075942.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 AK075961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353386 BC056164.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 416833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 96490 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119057 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 NM_153152.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 AK075942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 AK075961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 BC056164.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 NM_153152.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 AK075942.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 AK075961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353386 BC056164.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31257277 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119057 - -
          Whole Mouse Genome G4122A A_51_P353386 NM_153152.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 AK075942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 AK075961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 BC056164.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 NM_153152.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 AK075942.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 AK075961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P353386 BC056164.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139475 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139890 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13290089 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13459628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13340773 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13290094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13340769 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13340765 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13338984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13340770 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13340774 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13338956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13290086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257263 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257265 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31257219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31257198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13527899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13527908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13527913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13459628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13527919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13527933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257248 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13570324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257251 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257245 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257261 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257270 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31257228 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10452113 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452112 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452115 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452111 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452114 - -
          Affymetrix Mouse Gene 1.0 ST Array 10452116 - -
          Affymetrix HT MG-430 PM Array Plate 1427909_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1427237_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10452110 - -
          Mouse Genome 430 2.0 Array 1427237_PM_at - -
          Mouse Genome 430 2.0 Array 1427237_at - -
          Mouse Genome 430 2.0 Array 1427909_PM_at - -
          Mouse Genome 430 2.0 Array 1427909_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224904_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 224904_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700021837.mm.1 NM_153152.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021837.mm.1 NM_153152.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021837.mm.1 TC1700002332.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021836.mm.1 NM_153152.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021836.mm.1 NM_153152.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021836.mm.1 TC1700002332.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1427909_at - -
          Affymetrix HT Mouse Genome 430A Array 1427909_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427237_at - -
          Affymetrix HT Mouse Genome 430A Array 1427237_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 412870 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840066 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230931 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230931 - -
          mouse-6 v1.1 (Illumina) scl0224904.1_72-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840066 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0224904.1_72-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1230931 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230931 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0224904.1_72-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline