Search
Search
| Gene Symbol: Ago4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153177.3 | NP_694817.2 | 2 - 3 | 437 | 59 | |
| GenBank mRNA | AB081474.1 | - | 2 - 3 | 182 | 59 | |
| AK030018.1 | - | 2 - 3 | 453 | 59 | ||
| BB616446.1 | - | 2 - 3 | 437 | 59 | ||
| BC096023.1 | - | 2 - 3 | 219 | 30607807 | 59 | |
Set Membership: |
|