Search
Search
| Gene Symbol: Ago1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153403.2 | NP_700452.2 | 2 - 3 | 212 | 75 | |
| GenBank mRNA | AB081471.1 | - | 2 - 3 | 212 | 75 | |
| BC129914.1 | - | 2 - 3 | 212 | 40111808 | 75 | |
| BC129915.1 | - | 2 - 3 | 208 | 75 | ||
Set Membership: |
|