Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463327_m1
          See other IL1F9 GE Assays ›
          Gene Symbol:
          Il1f9
          Gene Name:
          interleukin 1 family, member 9
          Gene Aliases:
          Il36g
          Chromosome Location:
          Chr.2: 24186476 - 24193567 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463327_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il1f9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153511.3 NP_705731.2 4 - 5 669 69
          GenBank mRNA AK081783.1 - 4 - 5 664 69
          AK156687.1 - 666 69
          AY071843.1 - 3 - 4 395 69
          BC116227.1 - 3 - 4 395 40091521 69
          BC116228.1 - 3 - 4 395 40091522 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000376.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il1f9

          Panther Classification:

          Molecular Function -

          interleukin superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cytokine production
          immune system process
          inflammatory response
          cytokine-mediated signaling pathway
          positive regulation of interleukin-6 production
          innate immune response
          negative regulation of myoblast differentiation
          negative regulation of myoblast fusion
          cytokine activity
          interleukin-1 receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071176 AY071843.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071176 NM_153511.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071176 AK081783.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762043 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463541 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 383979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 79748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 424118 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66433 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141465 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P143393 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071176 AY071843.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071176 NM_153511.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071176 AK081783.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408116 NM_153511.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P408116 AK081783.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064549 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064576 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21064605 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P143393 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141465 - -
          Whole Mouse Genome G4122A A_51_P408116 NM_153511.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P408116 AK081783.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414013 AK156687.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414013 AY071843.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414013 BC116227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414013 BC116228.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P414013 NM_153511.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137796 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141465 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P143393 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01177911 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01177909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01177905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02050473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01177907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02050469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02050478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01177912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02050470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21064594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064539 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02131107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064535 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01948724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02050472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01948723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02050477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064526 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02050476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01918973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064537 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02050466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01948721 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02196752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02016629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02196754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21064547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01177909 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469789 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469790 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469788 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469787 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469792 - -
          Affymetrix HT MG-430 PM Array Plate 1425958_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469786 - -
          Mouse Genome 430 2.0 Array 1425958_PM_at - -
          Mouse Genome 430 2.0 Array 1425958_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215257_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215257_at - -
          Affymetrix Mouse Clariom S Array PSR0200001872.mm.1 NM_153511.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001872.mm.1 NM_153511.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001872.mm.1 TC0200000376.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200001871.mm.1 NM_153511.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001871.mm.1 NM_153511.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200001871.mm.1 TC0200000376.mm.1 MTA TC
          Mouse Expression Set 430 1425958_PM_at - -
          Mouse Expression Set 430 1425958_at - -
          Mouse Genome 430A 2.0 Array 1425958_at - -
          Mouse Genome 430A 2.0 Array 1425958_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425958_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425958_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 340643 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4810273 - -
          Illumina mouse-6 v1.0 expression beadchip 4810273 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810273 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658704 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2658704 - -
          mouse-6 v1.1 (Illumina) scl21204.5.1_17-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810273 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21204.5.1_17-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810273 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2658704 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2658704 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline