Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463494_m1
          See other ZBTB8B GE Assays ›
          Gene Symbol:
          Zbtb8b
          Gene Name:
          zinc finger and BTB domain containing 8b
          Gene Aliases:
          AV220464, BC023839, BOZF1, Zbtb8
          Chromosome Location:
          Chr.4: 129425765 - 129446972 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463494_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zbtb8b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153541.3 NP_705769.2 3 - 4 1227 70
          GenBank mRNA AK132152.1 - 3 - 4 1265 70
          AK140914.1 - 3 - 4 1216 70
          AK163930.1 - 3 - 4 1228 70
          BC023839.1 - 3 - 4 1222 5344921 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003594.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zbtb8b

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2430367 AK163930.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2430367 BC023839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2430367 AK132152.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2430367 AK140914.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2430367 NM_153541.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 176532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306478 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 772245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 795960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 40869 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 712139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 89196 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152520 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2430367 AK163930.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2430367 BC023839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2430367 AK132152.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2430367 AK140914.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2430367 NM_153541.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23296634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05399590 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152520 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00986811 - -
          Whole Mouse Genome G4122A A_52_P369613 AK163930.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P369613 BC023839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P369613 AK132152.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P369613 AK140914.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P369613 NM_153541.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109969 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152520 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P00986811 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03501984 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03501990 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03561279 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03992499 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03501988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23296624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03230461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04074626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03501991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03899303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03883175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04062062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03732521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03833308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03899294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03992499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04079045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03833306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296551 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296642 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03449953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03213528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03251238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03230466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03213530 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03768246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04042236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23296646 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516601 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516600 - -
          Affymetrix HT MG-430 PM Array Plate 1455193_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516596 - -
          Mouse Genome 430 2.0 Array 1455193_PM_at - -
          Mouse Genome 430 2.0 Array 1455193_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215627_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 215627_at - -
          Affymetrix Mouse Clariom S Array PSR0400029294.mm.1 TC0400003594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029294.mm.1 NM_153541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029294.mm.1 NM_153541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029291.mm.1 TC0400003594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029291.mm.1 NM_153541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029291.mm.1 NM_153541.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029293.mm.1 TC0400003594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029293.mm.1 NM_153541.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029293.mm.1 NM_153541.3 Refseq
          Mouse Expression Set 430 1455193_at - -
          Mouse Expression Set 430 1455193_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455193_at - -
          Affymetrix HT Mouse Genome 430B Array 1455193_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 712733 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6110280 - -
          Illumina mouse-6 v1.0 expression beadchip 6110280 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006110280 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2418483 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2418483 - -
          mouse-6 v1.1 (Illumina) scl23782.4.1_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 6110280 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23782.4.1_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6110280 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2418483 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2418483 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline