Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463571_m1
          See other GNL3 GE Assays ›
          Gene Symbol:
          Gnl3
          Gene Name:
          guanine nucleotide binding protein-like 3 (nucleolar)
          Gene Aliases:
          Ns
          Chromosome Location:
          Chr.14: 31012433 - 31019131 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463571_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnl3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153547.6 NP_705775.2 13 - 14 1657 80
          NR_104298.1 13 - 14 1609 80
          GenBank mRNA AA041888.1 - 228 80
          AK077523.1 - 13 - 14 1588 80
          AK167183.1 - 13 - 14 1541 80
          AY181025.1 - 13 - 14 1515 80
          BC018560.1 - 13 - 14 1496 80
          BC037996.1 - 13 - 14 1517 5052858 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001665.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Estrogen Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnl3

          Gene Ontology Categories:

          Function(s) Process(es)

          cell proliferation
          regulation of cell proliferation
          ribosome biogenesis
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          nucleotide binding
          GTPase activity
          protein binding
          GTP binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AY185498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 BC018560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 BC037996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AK167183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AY181025.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AY185498.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 BC018560.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 BC037996.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AK167183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P377376 AY181025.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637267 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 134372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 651094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136317 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164733 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149258 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AY185498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 BC018560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 BC037996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AK167183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AY181025.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AY185498.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 BC018560.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 BC037996.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AK167183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P377376 AY181025.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29509224 - -
          Whole Mouse Genome G4122A A_51_P377376 AY185498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 BC018560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 BC037996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 AK167183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 AY181025.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 AY185498.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 BC018560.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 BC037996.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 AK167183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P377376 AY181025.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111160 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29509156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509151 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11471708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11812743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29509172 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418485 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418490 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418491 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418487 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418493 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418484 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418492 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418496 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418495 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418486 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418494 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418497 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418489 - -
          Affymetrix HT MG-430 PM Array Plate 1433656_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418480 - -
          Mouse Genome 430 2.0 Array 1440766_at - -
          Mouse Genome 430 2.0 Array 1440766_PM_at - -
          Mouse Genome 430 2.0 Array 1433656_a_at - -
          Mouse Genome 430 2.0 Array 1433656_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30877_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 30877_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400012633.mm.1 NM_153547.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012633.mm.1 NR_104298.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012633.mm.1 TC1400001665.mm.1 MTA TC
          Mouse Expression Set 430 1433656_a_at - -
          Mouse Expression Set 430 1433656_PM_a_at - -
          Murine Genome U74v2 Set 98948_PM_at - -
          Murine Genome U74v2 Set 98948_at - -
          Mouse Genome 430A 2.0 Array 1433656_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1433656_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1433656_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1433656_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4810500 - -
          MouseWG-6 v2 Expression BeadChip 4050338 - -
          Illumina mouse-6 v1.0 expression beadchip 4050338 - -
          Illumina mouse-6 v1.0 expression beadchip 4810500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810500 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004050338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601639 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601639 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673441 - -
          mouse-6 v1.1 (Illumina) scl000383.1_1-S - -
          mouse-6 v1.1 (Illumina) scl45770.11.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4810500 - -
          Illumina Sentrix mouse-6.v1 bead array 4050338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000383.1_1-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45770.11.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810500 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4050338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3097726 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601639 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2673441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2673441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3097726 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601639 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45770.11.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000383.1_1-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline