Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463794_m1
          See other SYNE4 GE Assays ›
          Gene Symbol:
          Syne4
          Gene Name:
          spectrin repeat containing, nuclear envelope family member 4
          Gene Aliases:
          0610012K07Rik, AI428936, Nesp4
          Chromosome Location:
          Chr.7: 30314816 - 30319045 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463794_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Syne4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290565.1 NP_001277494.1 2 - 3 408 83
          NM_153577.2 NP_705805.1 2 - 3 408 83
          GenBank mRNA BC004761.1 - 2 - 3 357 3155758 83
          BC023803.1 - 2 - 3 395 5352953 83
          BC056649.1 - 2 - 3 347 5036575 83
          BE626968.1 - 2 - 3 333 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000559.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Syne4

          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of epithelial cell apical/basal polarity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 BC023803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 BG063039.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 NM_153577.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 AK002612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 BC023803.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 BG063039.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 NM_153577.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P147803 AK002612.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660166 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79938 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145965 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 BC023803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 BG063039.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 NM_153577.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 AK002612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 BC023803.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 BG063039.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 NM_153577.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P147803 AK002612.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25030934 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145965 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06154250 - -
          Whole Mouse Genome G4122A A_52_P147803 BC023803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 BG063039.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 NM_153577.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 AK002612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 BC023803.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 BG063039.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 NM_153577.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P147803 AK002612.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127678 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145965 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06582224 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06582227 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06582229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25030938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25031021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06020328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06582224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06464102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06466446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06027567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06464106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06390431 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06582228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06000439 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25030940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06464103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551875 - -
          Affymetrix HT MG-430 PM Array Plate 1427176_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1427175_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10551872 - -
          Mouse Genome 430 2.0 Array 1427176_s_at - -
          Mouse Genome 430 2.0 Array 1427175_at - -
          Mouse Genome 430 2.0 Array 1427176_PM_s_at - -
          Mouse Genome 430 2.0 Array 1427175_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233066_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233066_at - -
          Affymetrix Mouse Clariom S Array PSR0700004830.mm.1 TC0700000559.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004830.mm.1 NM_153577.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004830.mm.1 NM_153577.2 Refseq
          Mouse Expression Set 430 1427176_PM_s_at - -
          Mouse Expression Set 430 1427175_PM_at - -
          Mouse Expression Set 430 1427176_s_at - -
          Mouse Expression Set 430 1427175_at - -
          Murine Genome U74v2 Set 161008_PM_at - -
          Murine Genome U74v2 Set 161008_at - -
          Murine Genome U74v2 Set 161857_PM_r_at - -
          Murine Genome U74v2 Set 161857_r_at - -
          Mouse Genome 430A 2.0 Array 1427175_at - -
          Mouse Genome 430A 2.0 Array 1427175_PM_at - -
          Mouse Genome 430A 2.0 Array 1427176_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427176_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427176_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427176_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427175_at - -
          Affymetrix HT Mouse Genome 430A Array 1427175_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 532301 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2650433 - -
          Illumina mouse-6 v1.0 expression beadchip 2650433 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002650433 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2619639 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2619639 - -
          mouse-6 v1.1 (Illumina) scl32818.7.1_108-S - -
          Illumina Sentrix mouse-6.v1 bead array 2650433 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32818.7.1_108-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2650433 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2619639 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2619639 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32818.7.1_108-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline