Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463819_m1
          See other ATG4D GE Assays ›
          Gene Symbol:
          Atg4d
          Gene Name:
          autophagy related 4D, cysteine peptidase
          Gene Aliases:
          9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4
          Chromosome Location:
          Chr.9: 21265285 - 21274837 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463819_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atg4d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153583.10 NP_705811.8 1 - 2 433 72
          GenBank mRNA AK036282.1 - 1 - 2 420 72
          AK036571.1 - 1 - 2 422 72
          AK081002.1 - 1 - 2 420 72
          AK081605.1 - 1 - 2 411 72
          AK087258.1 - 1 - 2 423 72
          AK154130.1 - 1 - 2 426 72
          BC030861.1 - 1 - 2 410 4208700 72
          BC069851.1 - 1 - 2 385 6490007 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000191.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atg4d

          Panther Classification:

          Molecular Function -

          cysteine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          autophagosome assembly
          mitophagy
          C-terminal protein lipidation
          proteolysis
          protein targeting to membrane
          transport
          autophagy
          apoptotic process
          cellular response to nitrogen starvation
          protein transport
          protein processing
          nucleophagy
          protein delipidation
          cysteine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346228 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346227 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961135 BC030861.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961135 BC069851.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961135 AK154130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961135 AK215761.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1961135 AK036571.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 536475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 510176 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51064 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162938 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150466 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961135 BC030861.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961135 BC069851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961135 AK154130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961135 AK215761.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1961135 AK036571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P207106 AK044798.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P207106 AK080353.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P418624 AK036571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P418624 NM_153583.9 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P418624 AK036282.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26346228 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06718200 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01881824 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150466 - -
          Whole Mouse Genome G4122A A_52_P207106 AK044798.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P207106 AK080353.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418624 AK036571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418624 NM_153583.9 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P418624 AK036282.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526750 BC030861.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526750 BC069851.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526750 AK081605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526750 AK087258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P526750 AK154130.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121146 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114433 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150466 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08037590 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08037587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08037584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08037582 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346229 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346225 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346226 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346230 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26346228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26346238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07494687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07743424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08037582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07939390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08037584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07459146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07844674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07865022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07647106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07550368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07657739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08107805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26346214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08011366 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10583580 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583584 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583582 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583583 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583579 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583575 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583578 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583581 - -
          Affymetrix Mouse Gene 1.0 ST Array 10583576 - -
          Affymetrix HT MG-430 PM Array Plate 1439130_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433918_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591497 - -
          Affymetrix Mouse Gene 1.1 ST Array 10583573 - -
          Mouse Genome 430 2.0 Array 1439130_at - -
          Mouse Genome 430 2.0 Array 1433918_PM_at - -
          Mouse Genome 430 2.0 Array 1439130_PM_at - -
          Mouse Genome 430 2.0 Array 1433918_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235040_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 235040_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900001277.mm.1 NM_153583.10 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001277.mm.1 TC0900000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001279.mm.1 NM_153583.10 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001279.mm.1 TC0900000191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900001278.mm.1 NM_153583.10 Refseq
          Affymetrix Mouse Clariom S Array PSR0900001278.mm.1 TC0900000191.mm.1 MTA TC
          Mouse Expression Set 430 1433918_PM_at - -
          Mouse Expression Set 430 1433918_at - -
          Mouse Expression Set 430 1439130_PM_at - -
          Mouse Expression Set 430 1439130_at - -
          Mouse Genome 430A 2.0 Array 1433918_at - -
          Mouse Genome 430A 2.0 Array 1433918_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433918_at - -
          Affymetrix HT Mouse Genome 430A Array 1433918_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439130_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439130_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 434918 - -
          ABI Mouse Genome Survey Microarray 929561 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3290176 - -
          MouseWG-6 v2 Expression BeadChip 106840673 - -
          Illumina mouse-6 v1.0 expression beadchip 101500400 - -
          Illumina mouse-6 v1.0 expression beadchip 3290176 - -
          Illumina mouse-6 v1.0 expression beadchip 106650100 - -
          Illumina mouse-6 v1.0 expression beadchip 106400086 - -
          Illumina mouse-6 v1.0 expression beadchip 106840673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601710 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601710 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237020 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2449193 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2581779 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231100 - -
          mouse-6 v1.1 (Illumina) scl37228.10_481-S - -
          mouse-6 v1.1 (Illumina) scl00235040.2_157-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290176 - -
          Illumina Sentrix mouse-6.v1 bead array 106400086 - -
          Illumina Sentrix mouse-6.v1 bead array 106840673 - -
          Illumina Sentrix mouse-6.v1 bead array 106650100 - -
          Illumina Sentrix mouse-6.v1 bead array 101500400 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00235040.2_157-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106400086 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106650100 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101500400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290176 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106840673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601710 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601710 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237020 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231100 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2449193 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235412 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline