Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463932_m1
          See other TRPT1 GE Assays ›
          Gene Symbol:
          Trpt1
          Gene Name:
          tRNA phosphotransferase 1
          Gene Aliases:
          AU079016, AW045591, Tpt1, Tpt1h
          Chromosome Location:
          Chr.19: 6996131 - 6999048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463932_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153597.2 NP_705825.2 2 - 3 476 95
          GenBank mRNA BC027575.1 - 2 - 3 274 1364225 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000128.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpt1

          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA splicing, via endonucleolytic cleavage and ligation
          tRNA processing
          regulation of protein kinase activity
          tRNA 2'-phosphotransferase activity
          2'-phosphotransferase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 BC027575.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK141582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK048558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK090183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 NM_153597.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461183 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 BC027575.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK141582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK048558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK090183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 NM_153597.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 BC027575.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK141582.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK048558.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK090183.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 NM_153597.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937224 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148301 - -
          Whole Mouse Genome G4122A A_51_P337883 AK048558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 BC027575.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK141582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK048558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK090183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 NM_153597.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128150 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14306813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14306818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937223 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461034 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461036 - -
          Affymetrix HT MG-430 PM Array Plate 1452615_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461028 - -
          Mouse Genome 430 2.0 Array 1452615_s_at - -
          Mouse Genome 430 2.0 Array 1452615_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107328_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107328_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900001727.mm.1 NM_153597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001727.mm.1 NM_153597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001727.mm.1 TC1900000128.mm.1 MTA TC
          Mouse Expression Set 430 1452615_s_at - -
          Mouse Expression Set 430 1452615_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452615_s_at - -
          Mouse Genome 430A 2.0 Array 1452615_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452615_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452615_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 378453 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910093 - -
          Illumina mouse-6 v1.0 expression beadchip 5910093 - -
          Illumina mouse-6 v1.0 expression beadchip 6220056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910093 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2480638 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2439751 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2480638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2439751 - -
          mouse-6 v1.1 (Illumina) scl0107328.6_14-S - -
          mouse-6 v1.1 (Illumina) scl000496.1_7-S - -
          Illumina Sentrix mouse-6.v1 bead array 5910093 - -
          Illumina Sentrix mouse-6.v1 bead array 6220056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0107328.6_14-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000496.1_7-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2885981 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2885982 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2480638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2480638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2885981 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2885982 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0107328.6_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0