Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00463997_m1
          See other PRKAG3 GE Assays ›
          Gene Symbol:
          Prkag3
          Gene Name:
          protein kinase, AMP-activated, gamma 3 non-catatlytic subunit
          Gene Aliases:
          AMPKg3L, AMPKg3S, Amkg3, Ampkg3
          Chromosome Location:
          Chr.1: 74738922 - 74749590 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00463997_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkag3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153744.3 NP_714966.1 10 - 11 1276 64
          GenBank mRNA AF525500.1 - 10 - 11 1183 64
          AF525501.1 - 9 - 10 1204 64
          AK036585.1 - 10 - 11 1277 64
          BC116749.1 - 9 - 10 1140 40126068 64
          BC116777.1 - 9 - 10 1140 40126096 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002589.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkag3

          Panther Classification:

          Molecular Function -

          kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          glycogen biosynthetic process
          protein phosphorylation
          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          glucose transport
          nucleotide binding
          AMP-activated protein kinase activity
          ATP binding
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20350235 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077681 AF525500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077681 AF525501.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077681 AK036585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2077681 NM_153744.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 475371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567858 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 678987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 35315 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077681 AF525500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077681 AF525501.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077681 AK036585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2077681 NM_153744.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P107923 BC116749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P107923 BC116777.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P107923 AF525500.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P107923 AF525501.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P107923 AK036585.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20350258 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04148685 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00104726 - -
          Whole Mouse Genome G4122A A_52_P107923 BC116749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P107923 BC116777.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P107923 AF525500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P107923 AF525501.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P107923 AK036585.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P139398 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00882430 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00882440 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00882439 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00882427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00882441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20350235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20350265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00369858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00168587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00369860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00882436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00882442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350256 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00677767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01073424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01073425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00882438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00249417 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00905054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00905055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00905056 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01073426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20350252 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10355643 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355642 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355649 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355645 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355640 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355646 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355650 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355651 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355652 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355644 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355648 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355641 - -
          Affymetrix Mouse Gene 1.0 ST Array 10355653 - -
          Affymetrix HT MG-430 PM Array Plate 1444480_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10355639 - -
          Mouse Genome 430 2.0 Array 1444480_PM_at - -
          Mouse Genome 430 2.0 Array 1444480_at - -
          Affymetrix Mouse Clariom S Array PSR0100020944.mm.1 NM_153744.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020944.mm.1 NM_153744.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100020944.mm.1 TC0100002589.mm.1 MTA TC
          Mouse Expression Set 430 1444480_at - -
          Mouse Expression Set 430 1444480_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 548129 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106510735 - -
          Illumina mouse-6 v1.0 expression beadchip 106510735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2702917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2468772 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2702917 - -
          mouse-6 v1.1 (Illumina) scl16610.12.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 106510735 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106510735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3092923 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2702917 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2468772 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline