Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00464033_m1
          See other CTU2 GE Assays ›
          Gene Symbol:
          Ctu2
          Gene Name:
          cytosolic thiouridylase subunit 2 homolog (S. pombe)
          Gene Aliases:
          2310061F22Rik, C16orf84, Ncs2
          Chromosome Location:
          Chr.8: 122476137 - 122483092 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00464033_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ctu2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153775.2 NP_722470.2 4 - 5 367 64
          GenBank mRNA AK012440.1 - 4 - 5 333 64
          AK090391.1 - 302 64
          AK154995.1 - 4 - 5 302 64
          AK172353.1 - 2 - 3 810 64
          BC036332.1 - 4 - 5 320 3156706 64
          CJ067802.1 - 4 - 5 368 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001490.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ctu2

          Panther Classification:

          Molecular Function -

          RNA metabolism protein RNA processing factor nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA wobble uridine modification
          tRNA processing
          biological_process
          tRNA thio-modification
          tRNA binding
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966538 BC036332.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966538 AK090349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966538 AK090391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966538 AK172353.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966538 NM_153775.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027634 AK129095.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027634 AK079317.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027634 AK090349.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027634 AK090391.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027634 NM_153775.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500379 AK085074.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500379 AK141954.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500379 BC034531.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500379 NM_153543.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 824428 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 454123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476543 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131619 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966538 BC036332.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966538 AK090349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966538 AK090391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966538 AK172353.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966538 NM_153775.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027634 AK129095.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027634 AK079317.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027634 AK090349.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027634 AK090391.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027634 NM_153775.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26233664 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131619 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06669602 - -
          Whole Mouse Genome G4122A A_51_P133575 BC036332.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133575 AK090349.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133575 AK090391.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133575 AK154995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P133575 AK172353.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06935647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26233761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07300603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06935648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233674 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233693 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07262367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07300588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233684 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07413784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07300579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233644 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06925987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07300592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06925976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07300598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26233762 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576130 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576128 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576133 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576127 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576132 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576135 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576134 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576136 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576139 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576129 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576137 - -
          Affymetrix HT MG-430 PM Array Plate 1428350_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576124 - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66965_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66965_at - -
          Affymetrix Mouse Clariom S Array PSR0800012132.mm.1 NM_153775.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012132.mm.1 NM_153775.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012132.mm.1 TC0800001490.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800012133.mm.1 NM_153775.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012133.mm.1 NM_153775.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800012133.mm.1 TC0800001490.mm.1 MTA TC
          Mouse Expression Set 430 1428350_PM_at - -
          Mouse Expression Set 430 1439387_x_at - -
          Mouse Expression Set 430 1428350_at - -
          Mouse Expression Set 430 1439387_PM_x_at - -
          Murine Genome U74v2 Set 170002_PM_at - -
          Murine Genome U74v2 Set 170002_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 428704 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106660735 - -
          Illumina mouse-6 v1.0 expression beadchip 101990050 - -
          Illumina mouse-6 v1.0 expression beadchip 106660735 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1221168 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222859 - -
          Illumina Sentrix mouse-6.v1 bead array 106660735 - -
          Illumina Sentrix mouse-6.v1 bead array 101990050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101990050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106660735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2860725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2860725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212964 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222859 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline