Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00464101_m1
          See other CCDC64B GE Assays ›
          Gene Symbol:
          Ccdc64b
          Gene Name:
          coiled-coil domain containing 64B
          Gene Aliases:
          BICDR-2
          Chromosome Location:
          Chr.17: 23660523 - 23668818 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00464101_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccdc64b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153784.2 NP_722479.1 2 - 3 330 61
          GenBank mRNA AK036793.1 - 2 - 3 345 61
          AK133729.1 - 2 - 3 347 61
          BC038613.1 - 2 - 3 356 4458084 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000314.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccdc64b

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          Rab GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092908 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 BC038613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 NM_153784.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 AK036793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 AK133729.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 BC038613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 NM_153784.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 AK036793.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P471362 AK133729.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45554 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 185897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654855 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 120797 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148945 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 BC038613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 NM_153784.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 AK036793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 AK133729.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 BC038613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 NM_153784.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 AK036793.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P471362 AK133729.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31092923 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148945 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03292125 - -
          Whole Mouse Genome G4122A A_51_P471362 BC038613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 NM_153784.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 AK036793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 AK133729.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 BC038613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 NM_153784.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 AK036793.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P471362 AK133729.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120673 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132320 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148945 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13113753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13113760 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13113743 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31092908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31092881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13339071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13357139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13121802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13266553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092838 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13063779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13339061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490114 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13320582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13320584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474504 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13451509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13113753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13339055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31092881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13344494 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442315 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442320 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442314 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442316 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442317 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442318 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442319 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442312 - -
          Affymetrix HT MG-430 PM Array Plate 1460051_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442311 - -
          Mouse Genome 430 2.0 Array 1460051_at - -
          Mouse Genome 430 2.0 Array 1460051_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212733_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212733_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002277.mm.1 NM_153784.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002277.mm.1 TC1700000314.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002278.mm.1 NM_153784.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002278.mm.1 TC1700000314.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002276.mm.1 NM_153784.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002276.mm.1 TC1700000314.mm.1 MTA TC
          Mouse Expression Set 430 1460051_at - -
          Mouse Expression Set 430 1460051_PM_at - -
          Murine Genome U74v2 Set 137733_at - -
          Murine Genome U74v2 Set 137733_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1460051_at - -
          Affymetrix HT Mouse Genome 430B Array 1460051_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 876650 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4590358 - -
          Illumina mouse-6 v1.0 expression beadchip 4590358 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004590358 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258172 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258172 - -
          mouse-6 v1.1 (Illumina) scl51036.12.1_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 4590358 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51036.12.1_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4590358 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2849305 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2849305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline