| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796720 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796716 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796718 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796719 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_65_P06872 |
AK162396.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_65_P06872 |
AK142190.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_65_P06872 |
AK147662.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_65_P06872 |
AK044238.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_65_P06872 |
NM_153794.4 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
383459 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
590166 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
652810 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
774914 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
244605 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
452255 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611293 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
AK051173.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
AK079948.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
AK083688.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
NM_010466.1 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
X07439.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
932159 |
X07646.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
887552 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
840372 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
464723 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
934747 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
472743 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
593746 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
262477 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
533867 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
332054 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
394388 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
133582 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
662600 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
780283 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
532064 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
719340 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
121365 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
57194 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
105374 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
45344 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P116090 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P145354 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123175 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_65_P06872 |
AK162396.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_65_P06872 |
AK142190.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_65_P06872 |
AK147662.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_65_P06872 |
AK044238.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_65_P06872 |
NM_153794.4 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796689 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796707 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796696 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796694 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796702 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796690 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796716 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796710 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796720 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796715 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796699 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796717 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796704 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796686 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796692 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796705 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796712 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796700 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P31796713 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09038638 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P145354 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P09038619 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P116090 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P416727 |
BC095936.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P416727 |
AK162396.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P416727 |
BC038695.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P416727 |
AK142190.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P416727 |
AK147662.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P216313 |
AK162396.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P216313 |
AK142190.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P216313 |
AK147662.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P216313 |
AK044238.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P216313 |
NM_153794.4 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P101595 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P145354 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P116090 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14045975 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14045963 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P14045965 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796720 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796716 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796719 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P31796718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796495 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796506 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796604 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796614 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796557 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796660 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796632 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796487 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796583 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796594 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796560 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796482 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796602 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796518 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796570 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796575 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796537 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796640 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796501 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796627 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796650 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796664 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796674 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796648 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796488 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796511 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796546 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796554 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796566 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796694 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796491 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796631 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796633 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796573 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796605 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796704 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796507 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796550 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796538 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796504 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796484 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796522 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796641 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796493 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796502 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796586 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796636 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796505 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796569 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796577 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796531 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796683 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796601 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796524 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796663 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796635 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796591 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796689 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796578 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796503 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796553 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796589 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796510 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796625 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796516 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796657 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796717 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796607 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796519 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796497 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796700 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796686 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796597 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796527 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796508 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796564 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796610 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796599 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796621 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796563 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796615 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796612 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796533 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796543 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796673 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796549 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796574 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796697 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796547 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796514 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796706 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796498 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796624 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796517 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796520 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796643 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796593 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796544 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796720 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796681 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796669 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796540 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796535 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796616 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796659 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796521 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796666 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P31796552 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796699 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796696 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796691 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796680 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796678 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13899294 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13989950 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14065961 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14065944 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796502 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796706 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796712 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13711882 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796503 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796700 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796676 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796697 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796679 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796683 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796498 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796677 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796497 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796715 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14065947 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796689 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796710 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796504 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796681 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796690 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796705 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796673 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796493 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796704 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796694 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P13615859 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796491 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796501 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P14045975 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796713 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P31796484 |
- |
- |