Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00465044_m1
          See other MRPL40 GE Assays ›
          Gene Symbol:
          Mrpl40
          Gene Name:
          mitochondrial ribosomal protein L40
          Gene Aliases:
          Nlvcf
          Chromosome Location:
          Chr.16: 18872018 - 18876637 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00465044_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl40
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010922.2 NP_035052.2 2 - 3 256 88
          GenBank mRNA AA240860.1 - 2 - 3 253 88
          AF034092.1 - 2 - 3 256 88
          AK145885.1 - 2 - 3 254 88
          BC028436.1 - 2 - 3 221 1246513 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600001322.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl40

          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025665 AF034092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025665 AK018757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025665 AK145885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025665 BC028436.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2025665 NM_010922.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138529 AF034092.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138529 AK018757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138529 AK145885.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2138529 NM_010922.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248022 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 80385 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148570 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138529 AF034092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138529 AK018757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138529 AK145885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2138529 NM_010922.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025665 AF034092.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025665 AK018757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025665 AK145885.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025665 BC028436.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2025665 NM_010922.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P420630 AF034092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P420627 AF034092.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P278532 AK037846.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P278532 NR_033526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278532 AK037846.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P278532 NR_033526.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420630 AF034092.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P420627 AF034092.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125031 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12928779 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438403 - -
          Affymetrix HT MG-430 PM Array Plate 1448849_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10438400 - -
          Mouse Genome 430 2.0 Array 1448849_PM_at - -
          Mouse Genome 430 2.0 Array 1448849_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18100_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18100_at - -
          Affymetrix Mouse Clariom S Array PSR1600010923.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010923.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010923.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010927.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010927.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010927.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010926.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010926.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010926.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010929.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010929.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010929.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010931.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010931.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010931.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010925.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010925.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010925.mm.1 TC1600001322.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600010930.mm.1 NM_010922.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010930.mm.1 NM_010922.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1600010930.mm.1 TC1600001322.mm.1 MTA TC
          Mouse Expression Set 430 1448849_PM_at - -
          Mouse Expression Set 430 1448849_at - -
          Murine Genome U74v2 Set 93603_at - -
          Murine Genome U74v2 Set 93603_PM_at - -
          Mouse Genome 430A 2.0 Array 1448849_at - -
          Mouse Genome 430A 2.0 Array 1448849_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448849_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448849_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 350828 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2370215 - -
          Illumina mouse-6 v1.0 expression beadchip 2370215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002370215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609087 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609087 - -
          mouse-6 v1.1 (Illumina) scl48676.4_117-S - -
          Illumina Sentrix mouse-6.v1 bead array 2370215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl48676.4_117-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2370215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2609087 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2609087 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl48676.4_117-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline