Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00465150_m1
          See other PTPRS GE Assays ›
          Gene Symbol:
          Ptprs
          Gene Name:
          protein tyrosine phosphatase, receptor type, S
          Gene Aliases:
          AL022616, PTP, PTP-NU3, PTPNU-3, PTPsigma, Ptpt9, R-PTP-S, RPTPsigma
          Chromosome Location:
          Chr.17: 56412426 - 56476480 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00465150_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptprs
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252453.1 NP_001239382.1 2 - 3 332 62
          NM_001252455.1 NP_001239384.1 2 - 3 332 62
          NM_001252456.1 NP_001239385.1 2 - 3 309 62
          NM_011218.2 NP_035348.2 2 - 3 332 62
          GenBank mRNA AK032604.1 - 2 - 3 242 62
          AK163915.1 - 2 - 3 297 62
          BC052462.1 - 2 - 3 258 6834684 62
          BC054077.1 - 2 - 3 225 62
          BC083188.1 - 2 - 3 291 5709264 62
          CJ177868.1 - 2 - 3 335 62
          D28530.1 - 2 - 3 221 62
          D28531.1 - 2 - 3 221 62
          X82288.1 - 1 - 2 141 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002324.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptprs

          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          cell adhesion
          dephosphorylation
          spinal cord development
          cerebellum development
          hippocampus development
          cerebral cortex development
          corpus callosum development
          peptidyl-tyrosine dephosphorylation
          establishment of endothelial intestinal barrier
          phosphoprotein phosphatase activity
          protein tyrosine phosphatase activity
          protein binding
          hydrolase activity
          phosphatase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252453.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252453.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252455.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_001252456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_011218.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 NM_011218.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021747.mm.1 TC1700002324.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252453.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252453.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252455.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252455.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_001252456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_011218.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 NM_011218.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021748.mm.1 TC1700002324.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700021750.mm.1 NM_001252456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021750.mm.1 NM_001252456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700021750.mm.1 TC1700002324.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0