Search
Search

Kcnh2| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294162.1 | NP_001281091.1 | 5 - 6 | 1694 | 101 | |
| NM_013569.2 | NP_038597.2 | 9 - 10 | 2722 | 101 | ||
| GenBank mRNA | AF012868.1 | - | 9 - 10 | 2728 | 101 | |
| AF012869.1 | - | 5 - 6 | 1642 | 101 | ||
| AF034762.1 | - | 5 - 6 | 1686 | 101 | ||
| AK145127.1 | - | 1698 | 101 | |||
| AY374424.1 | - | 9 - 10 | 2407 | 101 | ||
| BC051016.1 | - | 9 - 10 | 2553 | 101 | ||
| BU707188.1 | - | 3 - 4 | 508 | 101 | ||
Set Membership: |
|