Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00465965_m1
          See other FBXO36 GE Assays ›
          Gene Symbol:
          Fbxo36
          Gene Name:
          F-box protein 36
          Gene Aliases:
          0610008D19Rik, 1110020F21Rik, 2410002G19Rik, D1Ertd757e
          Chromosome Location:
          Chr.1: 84839841 - 84900487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00465965_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo36
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025386.3 NP_079662.1 3 - 4 416 75
          GenBank mRNA AK002325.1 - 3 - 4 396 75
          AK003856.1 - 3 - 4 416 75
          AK008324.1 - 3 - 4 413 75
          AK010335.1 - 3 - 4 1229 75
          BC049581.1 - 3 - 4 393 6772363 75
          BY708252.1 - 3 - 4 413 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000725.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo36

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405975 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405974 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405972 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 BC049581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK010335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK002325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK003856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK008324.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 BC049581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK010335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK002325.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK003856.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P362959 AK008324.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 686260 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 585245 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722579 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 85449 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452503 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896578 AF153197.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896578 AK012319.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896578 AK041900.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896578 BC009136.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 896578 NM_011835.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 776033 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730615 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 494886 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 675735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545979 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489123 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 865444 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 938649 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483104 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 872240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193826 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 266553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 586791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628664 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553520 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782816 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 52598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615726 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 312677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 359183 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867959 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 31912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13761 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 54421 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 88498 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 160365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115863 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157198 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 BC049581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK010335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK002325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK003856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK008324.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 BC049581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK010335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK002325.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK003856.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P362959 AK008324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 BC049581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK010335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK002325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK003856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK008324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 BC049581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK010335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK002325.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK003856.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P362959 AK008324.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P167148 AK036072.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104066 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405974 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405973 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405972 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20405975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20405968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00140670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01077725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00140672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405986 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00140652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405995 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P32832592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20405993 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10347914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347912 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347911 - -
          Affymetrix HT MG-430 PM Array Plate 1419305_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1449418_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10347910 - -
          Mouse Genome 430 2.0 Array 1419305_a_at - -
          Mouse Genome 430 2.0 Array 1419305_PM_a_at - -
          Mouse Genome 430 2.0 Array 1449418_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449418_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66153_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66153_at - -
          Affymetrix Mouse Clariom S Array PSR0100006147.mm.1 NM_025386.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006147.mm.1 NM_025386.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100006147.mm.1 TC0100000725.mm.1 MTA TC
          Mouse Expression Set 430 1449418_PM_s_at - -
          Mouse Expression Set 430 1419305_a_at - -
          Mouse Expression Set 430 1419305_PM_a_at - -
          Mouse Expression Set 430 1449418_s_at - -
          Mouse Genome 430A 2.0 Array 1449418_s_at - -
          Mouse Genome 430A 2.0 Array 1419305_a_at - -
          Mouse Genome 430A 2.0 Array 1449418_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1419305_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419305_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449418_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1419305_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449418_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 910903 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1410086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717387 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2717387 - -
          mouse-6 v1.1 (Illumina) scl17733.6.1_38-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17733.6.1_38-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2717387 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2717387 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline