Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466042_m1
          See other MTFP1 GE Assays ›
          Gene Symbol:
          Mtfp1
          Gene Name:
          mitochondrial fission process 1
          Gene Aliases:
          1700020C11Rik, 2610507A21Rik, AV005788, Mtp18
          Chromosome Location:
          Chr.11: 4091481 - 4095431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466042_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mtfp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026443.4 NP_080719.2 3 - 4 571 85
          GenBank mRNA AK006148.1 - 3 - 4 492 85
          AK010971.1 - 3 - 4 528 85
          AK012058.1 - 3 - 4 538 85
          AK078381.1 - 3 - 4 572 85
          BC019205.1 - 3 - 4 581 85
          BC058737.1 - 3 - 4 521 5400435 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002106.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mtfp1

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial fission
          apoptotic process
          mitochondrial membrane organization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555607 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555608 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 BC019205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 BC058737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK078381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK012058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 BC019205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 BC058737.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK078381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK012058.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841150 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734037 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107561 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126808 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 BC019205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 BC058737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK078381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK012058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK216258.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 BC019205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 BC058737.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK078381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK012058.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 BC019205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 BC058737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK078381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK012058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 BC019205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 BC058737.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK078381.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK012058.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555589 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126808 - -
          Whole Mouse Genome G4122A A_51_P175699 BC019205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 BC058737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK078381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK012058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK216258.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 BC019205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 BC058737.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK078381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK012058.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P175699 AK216258.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128129 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126808 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09222658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555607 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555608 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09721020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555607 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09721024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555577 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09442122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09222659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09222665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555586 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09442130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09524503 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09222662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09442138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555596 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10383817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383818 - -
          Affymetrix HT MG-430 PM Array Plate 1424223_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10383813 - -
          Mouse Genome 430 2.0 Array 1424223_PM_at - -
          Mouse Genome 430 2.0 Array 1424223_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67900_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67900_at - -
          Affymetrix Mouse Clariom S Array PSR1100020438.mm.1 NM_026443.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020438.mm.1 NM_026443.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100020438.mm.1 TC1100002106.mm.1 MTA TC
          Affymetrix HT Mouse Genome 430A Array 1424223_at - -
          Affymetrix HT Mouse Genome 430A Array 1424223_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 582181 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3840438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003840438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2624145 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2624145 - -
          mouse-6 v1.1 (Illumina) scl40589.5.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 3840438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40589.5.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3840438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2624145 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2624145 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl40589.5.1_6-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline