Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466067_m1
          See other CNTD1 GE Assays ›
          Gene Symbol:
          Cntd1
          Gene Name:
          cyclin N-terminal domain containing 1
          Gene Aliases:
          1700051C09Rik, 4921513J16Rik, 5430417M23Rik, Cntd
          Chromosome Location:
          Chr.11: 101279203 - 101288701 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466067_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cntd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026562.2 NP_080838.1 4 - 5 687 72
          GenBank mRNA AK006754.1 - 4 - 5 473 72
          AK007245.1 - 4 - 5 688 72
          AK049116.1 - 4 - 5 795 72
          BC006866.1 - 4 - 5 746 3595113 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001647.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cntd1

          Gene Ontology Categories:

          Function(s) Process(es)

          reciprocal meiotic recombination
          spermatogenesis
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK049116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 BC006866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 NM_026562.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK007245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK032176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK049116.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 BC006866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 NM_026562.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK007245.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P467960 AK032176.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 883564 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 295157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664107 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13148 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160351 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK049116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 BC006866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 NM_026562.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK007245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK032176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK049116.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 BC006866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 NM_026562.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK007245.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P467960 AK032176.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28073057 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160351 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07466220 - -
          Whole Mouse Genome G4122A A_51_P467960 AK049116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 BC006866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 NM_026562.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 AK007245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 AK032176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 AK049116.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 BC006866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 NM_026562.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 AK007245.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467960 AK032176.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129592 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07466220 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09208581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09208564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28073071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09290800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08983628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09415911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09415908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09208581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09730862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09415925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09415919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28073053 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10381339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391412 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381343 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10391411 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10381342 - -
          Affymetrix HT MG-430 PM Array Plate 1439405_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10391410 - -
          Affymetrix Mouse Gene 1.1 ST Array 10381334 - -
          Mouse Genome 430 2.0 Array 1432480_PM_a_at - -
          Mouse Genome 430 2.0 Array 1439405_x_at - -
          Mouse Genome 430 2.0 Array 1432480_a_at - -
          Mouse Genome 430 2.0 Array 1439405_PM_x_at - -
          Mouse Genome 430 2.0 Array 1460686_at - -
          Mouse Genome 430 2.0 Array 1460686_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68107_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68107_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100015432.mm.1 NM_026562.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015432.mm.1 NM_026562.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015432.mm.1 TC1100001647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015427.mm.1 NM_026562.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015427.mm.1 NM_026562.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015427.mm.1 TC1100001647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015433.mm.1 NM_026562.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015433.mm.1 NM_026562.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015433.mm.1 TC1100001647.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015431.mm.1 NM_026562.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015431.mm.1 NM_026562.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015431.mm.1 TC1100001647.mm.1 MTA TC
          Mouse Expression Set 430 1432480_PM_a_at - -
          Mouse Expression Set 430 1427196_PM_at - -
          Mouse Expression Set 430 1427196_at - -
          Mouse Expression Set 430 1432480_a_at - -
          Mouse Expression Set 430 1460686_at - -
          Mouse Expression Set 430 1460686_PM_at - -
          Murine Genome U74v2 Set 132114_f_at - -
          Murine Genome U74v2 Set 132114_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1460686_at - -
          Mouse Genome 430A 2.0 Array 1427196_at - -
          Mouse Genome 430A 2.0 Array 1460686_PM_at - -
          Mouse Genome 430A 2.0 Array 1432480_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1432480_a_at - -
          Mouse Genome 430A 2.0 Array 1427196_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 884113 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 940632 - -
          Illumina mouse-6 v1.0 expression beadchip 940632 - -
          Illumina mouse-6 v1.0 expression beadchip 106620154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000940632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2609984 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701760 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2701760 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227270 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2609984 - -
          mouse-6 v1.1 (Illumina) scl068107.7_175-S - -
          Illumina Sentrix mouse-6.v1 bead array 940632 - -
          Illumina Sentrix mouse-6.v1 bead array 106620154 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl068107.7_175-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106620154 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 940632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3075270 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3154335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3075270 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701759 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2550252 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3154335 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline