Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466088_m1
          See other DEB1 GE Assays ›
          Gene Symbol:
          Deb1
          Gene Name:
          differentially expressed in B16F10 1
          Gene Aliases:
          1110020I04Rik, AI463209, Ss18l2
          Chromosome Location:
          Chr.9: 121710389 - 121712921 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466088_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Deb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026794.2 NP_081070.1 2 - 3 158 104
          GenBank mRNA AK003863.1 - 2 - 3 166 104
          AK078209.1 - 1 - 2 270 104
          AK213061.1 - 1 - 2 48 104
          BC027548.1 - 2 - 3 158 1532650 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001623.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Deb1

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885061 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885059 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088716 BC027548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088716 NM_026794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088716 AK078209.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088715 BC027548.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088715 NM_026794.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088715 AK003863.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2088715 AK049702.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 298349 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503626 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115570 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088716 BC027548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088716 NM_026794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088716 AK078209.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088715 BC027548.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088715 NM_026794.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088715 AK003863.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2088715 AK049702.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26885060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06952447 - -
          Whole Mouse Genome G4122A A_52_P432787 AK085962.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432787 AK213061.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432787 BC027548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432787 NM_026794.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432787 AK003863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119715 BC027548.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119715 NM_026794.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119715 AK003863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P119715 AK078209.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100269 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06952447 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08095872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08095874 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885058 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885060 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885059 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26885061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26884997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26884996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26884999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26885081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08095874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07464983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08095877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07861571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08155962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26885048 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10590384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590388 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10590387 - -
          Affymetrix HT MG-430 PM Array Plate 1427955_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10590383 - -
          Mouse Genome 430 2.0 Array 1427955_a_at - -
          Mouse Genome 430 2.0 Array 1427955_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26901_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 26901_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900013578.mm.1 NM_026794.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013578.mm.1 NM_026794.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013578.mm.1 TC0900001623.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900013580.mm.1 NM_026794.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013580.mm.1 NM_026794.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900013580.mm.1 TC0900001623.mm.1 MTA TC
          Murine Genome U74v2 Set 95478_at - -
          Murine Genome U74v2 Set 95478_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427955_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1427955_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 542048 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570300 - -
          Illumina mouse-6 v1.0 expression beadchip 4570300 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570300 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2652971 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2652971 - -
          mouse-6 v1.1 (Illumina) scl026901.3_41-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570300 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl026901.3_41-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570300 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2652971 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2652971 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl026901.3_41-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline