Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466126_m1
          See other SPSB3 GE Assays ›
          Gene Symbol:
          Spsb3
          Gene Name:
          splA/ryanodine receptor domain and SOCS box containing 3
          Gene Aliases:
          2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
          Chromosome Location:
          Chr.17: 24886349 - 24892147 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466126_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spsb3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163750.1 NP_001157222.1 2 - 3 565 77
          NM_001163751.1 NP_001157223.1 2 - 3 220 77
          NM_027141.2 NP_081417.2 2 - 3 241 77
          GenBank mRNA AF403038.1 - 2 - 3 241 77
          AK009325.1 - 2 - 3 241 77
          AK076170.1 - 2 - 3 517 77
          AK082768.1 - 2 - 3 164 77
          AK156579.1 - 2 - 3 503 77
          AK167474.1 - 2 - 3 481 77
          AK218464.1 - 1 - 2 49 77
          AK220234.1 - 2 - 3 503 77
          BC072556.1 - 2 - 3 526 5691114 77
          BY276751.1 - 2 - 3 221 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000356.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spsb3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          intracellular signal transduction
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965114 AK220234.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965114 BC026523.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965114 BC072556.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965114 AK167474.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965114 AK082768.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358197 AK004859.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358197 AK008084.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358197 AK028573.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358197 BC029194.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358197 NM_172395.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948457 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965114 AK220234.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965114 BC026523.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965114 BC072556.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965114 AK167474.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965114 AK082768.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100108 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31100124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08740822 - -
          Whole Mouse Genome G4122A A_52_P613418 AK220234.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P613418 BC026523.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P613418 BC072556.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P613418 AK167474.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P613418 AK206769.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13494917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13494922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31100103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13531989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13531990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13230893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31100103 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442631 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442635 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442636 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442634 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442630 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442632 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442633 - -
          Affymetrix Mouse Gene 1.0 ST Array 10442637 - -
          Affymetrix HT MG-430 PM Array Plate 1417891_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10442629 - -
          Mouse Genome 430 2.0 Array 1417891_PM_at - -
          Mouse Genome 430 2.0 Array 1417891_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79043_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 79043_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700002808.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002808.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002808.mm.1 NM_001163751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002808.mm.1 NM_001163751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002808.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_001163751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_001163751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_027141.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 NM_027141.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002814.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_001163751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_001163751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_027141.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 NM_027141.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002815.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002811.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002811.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002811.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_001163751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_001163751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_027141.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 NM_027141.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002821.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_001163750.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_001163750.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_001163751.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_001163751.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_027141.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 NM_027141.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002818.mm.1 TC1700000356.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700002813.mm.1 NM_027141.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002813.mm.1 NM_027141.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700002813.mm.1 TC1700000356.mm.1 MTA TC
          Mouse Expression Set 430 1417891_PM_at - -
          Mouse Expression Set 430 1417891_at - -
          Mouse Expression Set 430 1442853_at - -
          Mouse Expression Set 430 1442853_PM_at - -
          Mouse Expression Set 430 1460628_at - -
          Mouse Expression Set 430 1460628_PM_at - -
          Murine Genome U74v2 Set 137082_at - -
          Murine Genome U74v2 Set 137082_PM_at - -
          Mouse Genome 430A 2.0 Array 1417891_PM_at - -
          Mouse Genome 430A 2.0 Array 1417891_at - -
          Affymetrix HT Mouse Genome 430A Array 1417891_at - -
          Affymetrix HT Mouse Genome 430A Array 1417891_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1740239 - -
          Illumina mouse-6 v1.0 expression beadchip 1740239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001740239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2645276 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645276 - -
          mouse-6 v1.1 (Illumina) scl50999.7.56_77-S - -
          Illumina Sentrix mouse-6.v1 bead array 1740239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50999.7.56_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1740239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2994108 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645276 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2645276 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2994108 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50999.7.56_77-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline