Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466138_m1
          See other TMEM147 GE Assays ›
          Gene Symbol:
          Tmem147
          Gene Name:
          transmembrane protein 147
          Gene Aliases:
          2010004E11Rik, 5033425B17Rik, Nifie14
          Chromosome Location:
          Chr.7: 30727701 - 30729534 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466138_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem147
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027215.2 NP_081491.2 2 - 3 261 125
          GenBank mRNA AK007753.1 - 2 - 3 222 125
          AK008089.1 - 2 - 3 215 125
          AK010168.1 - 2 - 3 250 125
          BC035323.1 - 2 - 3 234 125
          BC147300.1 - 2 - 3 215 9056333 125
          BC147301.1 - 2 - 3 215 9056336 125

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002775.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem147

          Gene Ontology Categories:

          Function(s) Process(es)

          protein destabilization
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033087 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033084 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033083 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC147300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC147301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC046908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 NM_027215.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 AK007753.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC147300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC147301.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 BC046908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 NM_027215.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P523330 AK007753.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967771 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152270 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC147300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC147301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC046908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 NM_027215.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 AK007753.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC147300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC147301.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 BC046908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 NM_027215.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P523330 AK007753.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25033085 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152270 - -
          Whole Mouse Genome G4122A A_52_P523330 BC147300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 BC147301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 BC046908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 NM_027215.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 AK007753.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 BC147300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 BC147301.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 BC046908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 NM_027215.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P523330 AK007753.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118045 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152270 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312654 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06312627 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033088 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033087 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033084 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033086 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25033083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25032996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25033152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06312632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06312616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25033088 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562104 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562103 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562101 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562102 - -
          Affymetrix HT MG-430 PM Array Plate 1428465_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10562096 - -
          Mouse Genome 430 2.0 Array 1428465_PM_at - -
          Mouse Genome 430 2.0 Array 1428465_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69804_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69804_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700024777.mm.1 TC0700002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024777.mm.1 NM_027215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024777.mm.1 NM_027215.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024779.mm.1 TC0700002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024779.mm.1 NM_027215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024779.mm.1 NM_027215.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024778.mm.1 TC0700002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024778.mm.1 NM_027215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024778.mm.1 NM_027215.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024780.mm.1 TC0700002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700024780.mm.1 NM_027215.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700024780.mm.1 NM_027215.2 Refseq
          Mouse Expression Set 430 1428465_at - -
          Mouse Expression Set 430 1428465_PM_at - -
          Murine Genome U74v2 Set 160235_at - -
          Murine Genome U74v2 Set 160235_PM_at - -
          Mouse Genome 430A 2.0 Array 1428465_at - -
          Mouse Genome 430A 2.0 Array 1428465_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428465_at - -
          Affymetrix HT Mouse Genome 430A Array 1428465_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 911681 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3290577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003290577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775751 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775751 - -
          mouse-6 v1.1 (Illumina) scl31512.7.1_16-S - -
          Illumina Sentrix mouse-6.v1 bead array 3290577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31512.7.1_16-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3290577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2775751 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775751 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31512.7.1_16-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline