Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466180_m1
          See other TMEM70 GE Assays ›
          Gene Symbol:
          Tmem70
          Gene Name:
          transmembrane protein 70
          Gene Aliases:
          1110020A09Rik, 2210416J16Rik
          Chromosome Location:
          Chr.1: 16665191 - 16678275 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466180_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem70
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026392.1 NP_080668.1 2 - 3 422 89
          NM_027415.2 NP_081691.2 2 - 3 419 89
          GenBank mRNA AK003839.1 - 2 - 3 404 89
          AK005075.1 - 2 - 3 404 89
          AK008955.1 - 2 - 3 388 89
          AK151280.1 - 2 - 3 394 89
          AK167069.1 - 2 - 3 345 89
          AK169031.1 - 2 - 3 352 89
          BC011320.1 - 2 - 3 348 3979025 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000101.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem70

          Gene Ontology Categories:

          Function(s) Process(es)

          mitochondrial proton-transporting ATP synthase complex assembly
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056642 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056644 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK167069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK169031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK181541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 BC011320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK003839.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK167069.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK169031.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK181541.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 BC011320.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P259415 AK003839.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 203804 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 279680 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 245094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395686 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 967781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 419434 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 100026 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100214 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK167069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK169031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK181541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 BC011320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK003839.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK167069.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK169031.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK181541.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 BC011320.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P259415 AK003839.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20056648 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100214 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04024408 - -
          Whole Mouse Genome G4122A A_51_P259415 AK167069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK169031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK181541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 BC011320.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK003839.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK167069.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK169031.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK181541.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 BC011320.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P259415 AK003839.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120930 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100214 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04024408 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00417894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056646 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20056642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20056664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00730372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01039174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00597980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00857174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01039175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01130993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00610619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056640 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00878182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00039570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00698245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20056669 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10344964 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344965 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344963 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344962 - -
          Affymetrix Mouse Gene 1.0 ST Array 10344961 - -
          Affymetrix HT MG-430 PM Array Plate 1424541_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10344960 - -
          Mouse Genome 430 2.0 Array 1424541_PM_at - -
          Mouse Genome 430 2.0 Array 1424541_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70397_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70397_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100000744.mm.1 NM_026392.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000744.mm.1 NM_026392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000744.mm.1 NM_027415.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000744.mm.1 NM_027415.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000744.mm.1 TC0100000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100000743.mm.1 NM_026392.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000743.mm.1 NM_026392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000743.mm.1 NM_027415.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000743.mm.1 NM_027415.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000743.mm.1 TC0100000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100000746.mm.1 NM_026392.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000746.mm.1 NM_026392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000746.mm.1 NM_027415.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000746.mm.1 NM_027415.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000746.mm.1 TC0100000101.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100000745.mm.1 NM_026392.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000745.mm.1 NM_026392.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000745.mm.1 NM_027415.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000745.mm.1 NM_027415.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100000745.mm.1 TC0100000101.mm.1 MTA TC
          Mouse Expression Set 430 1424541_at - -
          Mouse Expression Set 430 1424541_PM_at - -
          Murine Genome U74v2 Set 94829_PM_at - -
          Murine Genome U74v2 Set 94829_at - -
          Mouse Genome 430A 2.0 Array 1424541_at - -
          Mouse Genome 430A 2.0 Array 1424541_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424541_at - -
          Affymetrix HT Mouse Genome 430A Array 1424541_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 853785 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090142 - -
          Illumina mouse-6 v1.0 expression beadchip 1090142 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090142 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2678389 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2678389 - -
          mouse-6 v1.1 (Illumina) scl070397.4_136-S - -
          Illumina Sentrix mouse-6.v1 bead array 1090142 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl070397.4_136-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090142 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2678389 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3153614 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3153614 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2678389 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline