| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2007457 |
NM_028801.2 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
20976 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217069 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
784284 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
214131 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
894206 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
29578 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
49100 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
92964 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
516391 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
367999 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465105 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839434 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
403101 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521029 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
278631 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82740 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305162 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
970047 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
832857 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
AK011628.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
AK144126.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
AK146696.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
AK146913.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
BC005453.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
BC049783.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355859 |
NM_008709.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
388108 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
130234 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
45519 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
28830 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P127523 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137941 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2007457 |
NM_028801.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P122071 |
NM_028801.2 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P122071 |
AF369933.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P122071 |
AJ311906.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P122071 |
AK009011.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P127523 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01656509 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P137941 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P06397825 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P01656553 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P122071 |
NM_028801.2 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P122071 |
AF369933.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P122071 |
AJ311906.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P122071 |
AK009011.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P121248 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P127523 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P137941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591186 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591106 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591241 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591136 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591236 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591345 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591359 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591131 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591405 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591259 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591291 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591406 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591232 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591351 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591472 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591417 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591276 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591478 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591396 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591377 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591390 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591387 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591479 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591293 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591272 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591183 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591174 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591453 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591280 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591457 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591227 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591180 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591140 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591171 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591447 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591218 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591111 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591243 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591192 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591213 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591270 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591460 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591209 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591423 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591327 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591348 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591382 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591329 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591219 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591145 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591202 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591385 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591374 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591459 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591424 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591436 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591147 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591123 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591452 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591273 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591437 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591228 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591226 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591414 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591199 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591197 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591407 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591400 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591214 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591247 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591455 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591172 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591184 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591269 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591456 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591170 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591261 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591109 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591401 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591262 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591322 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591379 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591205 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591427 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591367 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591268 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591143 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591222 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591133 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591398 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591168 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591451 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591224 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591283 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591135 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591163 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591234 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591465 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591354 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591403 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591212 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591220 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591426 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591179 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591444 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591127 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591362 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591383 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591257 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591433 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591450 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591134 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591125 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591190 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591449 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591121 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591157 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591388 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591196 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591176 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591119 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591149 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591440 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591281 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591116 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591153 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591204 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591357 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591364 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591216 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591263 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591381 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591238 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591421 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591282 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591249 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591477 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591195 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591419 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591254 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591150 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591287 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591394 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591118 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591324 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591252 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591139 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591265 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591100 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591391 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591412 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591430 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591475 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591425 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591467 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591230 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591355 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591159 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591360 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591470 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591274 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591126 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591410 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591408 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591112 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591279 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591431 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591370 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591240 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591271 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591231 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591138 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591207 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591188 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P25591203 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05972775 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591140 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05972776 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591174 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591116 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591103 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06005846 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591190 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591102 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591172 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591184 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06212268 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06005832 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591112 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591104 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591149 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591160 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591145 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591166 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591109 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591196 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591121 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591194 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591193 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06212270 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591147 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591106 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591170 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06216759 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591113 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591192 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591168 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591188 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591180 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591118 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591136 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06216749 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591156 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591195 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591131 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06005827 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591186 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591111 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591134 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591135 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591126 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05972777 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591129 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591150 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591161 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591163 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591199 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591143 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591153 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591176 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P25591197 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P05972781 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06659852 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P06216755 |
- |
- |