Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00466471_m1
          See other SLX1B GE Assays ›
          Gene Symbol:
          Slx1b
          Gene Name:
          SLX1 structure-specific endonuclease subunit homolog B (S. cerevisiae)
          Gene Aliases:
          1110030E23Rik, 2410170E21Rik, 4833422P03Rik, AI853643, Giyd1, Giyd2
          Chromosome Location:
          Chr.7: 126688927 - 126695783 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00466471_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slx1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029420.4 NP_083696.2 1 - 2 360 87
          GenBank mRNA AK010748.1 - 1 - 2 361 87
          AK010828.1 - 1 - 2 324 87
          AK014753.1 - 1 - 2 217 87
          AK049139.1 - 1 - 2 361 87
          BC055909.1 - 1 - 2 312 87
          BC145688.1 - 1 - 2 225 40130991 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004292.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slx1b

          Panther Classification:

          Molecular Function -

          endodeoxyribonuclease

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA metabolic process
          DNA repair
          DNA recombination
          cellular response to DNA damage stimulus
          nucleic acid phosphodiester bond hydrolysis
          nuclease activity
          endonuclease activity
          crossover junction endodeoxyribonuclease activity
          hydrolase activity
          5'-flap endonuclease activity
          phosphoric ester hydrolase activity
          T/G mismatch-specific endonuclease activity
          retroviral 3' processing activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501810 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501812 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC145688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC055909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC145468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 AK010828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 AK049139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC145688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC055909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 BC145468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 AK010828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P113773 AK049139.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097945 AK153779.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097945 AK163709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097945 NR_033446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097945 NM_029420.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890427 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582475 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468770 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107259 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097945 AK153779.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097945 AK163709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097945 NR_033446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097945 NM_029420.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC145688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC055909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC145468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 AK010828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 AK049139.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC145688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC055909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 BC145468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 AK010828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P113773 AK049139.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25501791 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01630161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107259 - -
          Whole Mouse Genome G4122A A_51_P113773 BC145688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 BC055909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 BC145468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 AK010828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 AK049139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 BC145688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 BC055909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 BC145468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 AK010828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P113773 AK049139.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335104 AK153779.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335104 AK163709.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335104 NR_033446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335104 AI853643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335104 AK003973.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131136 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107259 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06152121 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06152107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06390421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06155482 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501812 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501811 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25501810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25501754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501794 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06012284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501785 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06516411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06380165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501779 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501759 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06498398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06012290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06140146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06430285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501789 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25501767 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568021 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568020 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568015 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568022 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568016 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568019 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568023 - -
          Affymetrix HT MG-430 PM Array Plate 1460095_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1454082_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1455219_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568017 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568011 - -
          Mouse Genome 430 2.0 Array 1454082_PM_a_at - -
          Mouse Genome 430 2.0 Array 1454082_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75764_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 75764_at - -
          Affymetrix Mouse Clariom S Array PSR0700035889.mm.1 NM_029420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035889.mm.1 NM_029420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035889.mm.1 NR_033446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035889.mm.1 NR_033446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035889.mm.1 TC0700004292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035893.mm.1 NM_029420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035893.mm.1 NM_029420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035893.mm.1 NR_033446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035893.mm.1 NR_033446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035893.mm.1 TC0700004292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035891.mm.1 NM_029420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035891.mm.1 NM_029420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035891.mm.1 NR_033446.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035891.mm.1 NR_033446.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035891.mm.1 TC0700004292.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700035888.mm.1 NM_029420.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035888.mm.1 NM_029420.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700035888.mm.1 TC0700004292.mm.1 MTA TC
          Mouse Expression Set 430 1454082_a_at - -
          Mouse Expression Set 430 1454082_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1454082_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1454082_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454082_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454082_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924268 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105690333 - -
          MouseWG-6 v2 Expression BeadChip 104540026 - -
          Illumina mouse-6 v1.0 expression beadchip 104540026 - -
          Illumina mouse-6 v1.0 expression beadchip 105690333 - -
          Illumina mouse-6 v1.0 expression beadchip 460400 - -
          Illumina mouse-6 v1.0 expression beadchip 106370519 - -
          Illumina mouse-6 v1.0 expression beadchip 100110082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460400 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242863 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1257882 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2555803 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242863 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227684 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238519 - -
          mouse-6 v1.1 (Illumina) scl30676.4_510-S - -
          Illumina Sentrix mouse-6.v1 bead array 106370519 - -
          Illumina Sentrix mouse-6.v1 bead array 104540026 - -
          Illumina Sentrix mouse-6.v1 bead array 105690333 - -
          Illumina Sentrix mouse-6.v1 bead array 460400 - -
          Illumina Sentrix mouse-6.v1 bead array 100110082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30676.4_510-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105690333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104540026 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100110082 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460400 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106370519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2966991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2966991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2555803 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227684 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257882 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1238519 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732394 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30676.4_510-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline