Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00467618_m1
          See other GPRC6A GE Assays ›
          Gene Symbol:
          Gprc6a
          Gene Name:
          G protein-coupled receptor, family C, group 6, member A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 51614688 - 51631614 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00467618_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gprc6a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153071.1 NP_694711.1 3 - 4 1365 96
          GenBank mRNA AY101365.1 - 3 - 4 1365 96
          AY735399.1 - 3 - 4 1338 96
          BC108998.2 - 3 - 4 1365 40048019 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002148.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Ion Channel Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gprc6a

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          calcium-mediated signaling
          response to amino acid
          signal transducer activity
          G-protein coupled receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097645 AY101365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2097645 NM_153071.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164771 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324837 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 346791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 921732 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 576790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 973332 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131042 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931034 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731099 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693008 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150927 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 73636 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 28393 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34521 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 4984 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136191 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117536 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160926 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097645 AY101365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2097645 NM_153071.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339038 BC108998.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339038 AY735400.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339038 AY735401.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339038 AY101365.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P339038 AY735399.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27134094 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117536 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160926 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02123704 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136191 - -
          Whole Mouse Genome G4122A A_51_P339038 BC108998.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339038 AY735400.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339038 AY735401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339038 AY101365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P339038 AY735399.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130258 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160926 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136191 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117536 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08760618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08760626 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08760622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08760617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08760619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27134034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27133957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27133999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08355561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08616493 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27133996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27133992 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08760626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27133994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08856744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08272490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08516557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27133993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27134051 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10369037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10369034 - -
          Affymetrix HT MG-430 PM Array Plate 1428053_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10369028 - -
          Mouse Genome 430 2.0 Array 1428053_PM_at - -
          Mouse Genome 430 2.0 Array 1428053_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 210198_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 210198_at - -
          Affymetrix Mouse Clariom S Array PSR1000015057.mm.1 NM_153071.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015057.mm.1 NM_153071.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000015057.mm.1 TC1000002148.mm.1 MTA TC
          Mouse Expression Set 430 1428053_PM_at - -
          Mouse Expression Set 430 1428053_at - -
          Mouse Genome 430A 2.0 Array 1428053_at - -
          Mouse Genome 430A 2.0 Array 1428053_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1428053_at - -
          Affymetrix HT Mouse Genome 430A Array 1428053_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 577236 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4560576 - -
          Illumina mouse-6 v1.0 expression beadchip 4560576 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560576 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588955 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588955 - -
          mouse-6 v1.1 (Illumina) scl37976.6.1_275-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560576 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37976.6.1_275-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560576 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2588955 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588955 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline