Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00467928_m1
          See other PEO1 GE Assays ›
          Gene Symbol:
          Peo1
          Gene Name:
          progressive external ophthalmoplegia 1 (human)
          Gene Aliases:
          D19Ertd626e, PEO, Twinl
          Chromosome Location:
          Chr.19: 45006079 - 45012762 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00467928_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Peo1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153796.3 NP_722491.2 1 - 2 1815 74
          GenBank mRNA AK146244.1 - 1 - 2 1709 74
          AY059385.1 - 1 - 2 1277 74
          BC071195.1 - 1 - 2 1815 5100486 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000661.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Peo1

          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA replication
          mitochondrial DNA replication
          DNA unwinding involved in DNA replication
          transcription from mitochondrial promoter
          DNA duplex unwinding
          protein hexamerization
          protein homooligomerization
          nucleotide binding
          protease binding
          DNA helicase activity
          single-stranded DNA binding
          helicase activity
          ATP binding
          hydrolase activity
          5'-3' DNA helicase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32135822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32135821 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 BC034909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 BC071195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 AK146244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 NM_153796.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 AK054159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 BC034909.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 BC071195.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 AK146244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 NM_153796.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P495372 AK054159.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 AJ237585.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 AK033995.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 AK039180.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 AK043845.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 NM_178142.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 XM_001475371.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 358792 XM_485604.4 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 439278 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 954128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136632 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P151217 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 BC034909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 BC071195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 AK146244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 NM_153796.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 AK054159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 BC034909.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 BC071195.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 AK146244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 NM_153796.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P495372 AK054159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261585 BC034909.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261585 BC071195.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261585 AK146244.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261585 AY059385.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P261585 NM_153796.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32135863 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09182652 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P151217 - -
          Whole Mouse Genome G4122A A_51_P261585 BC034909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261585 BC071195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261585 AK146244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261585 AY059385.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P261585 NM_153796.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 BC034909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 BC071195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 AK146244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 NM_153796.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 AK054159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 BC034909.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 BC071195.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 AK146244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 NM_153796.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P495372 AK054159.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136038 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136916 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P151217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14265540 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14265528 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14265533 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32135822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32135821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32135895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135857 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14265534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135846 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14265540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14265525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135835 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14405044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14407661 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135854 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135848 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135840 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32135862 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463453 - -
          Affymetrix HT MG-430 PM Array Plate 1434168_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463448 - -
          Mouse Genome 430 2.0 Array 1434168_at - -
          Mouse Genome 430 2.0 Array 1434168_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226153_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226153_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900005845.mm.1 NM_153796.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005845.mm.1 TC1900000661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005847.mm.1 NM_153796.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005847.mm.1 TC1900000661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900005848.mm.1 NM_153796.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005848.mm.1 TC1900000661.mm.1 MTA TC
          Mouse Expression Set 430 1434168_PM_at - -
          Mouse Expression Set 430 1434168_at - -
          Affymetrix HT Mouse Genome 430B Array 1434168_at - -
          Affymetrix HT Mouse Genome 430B Array 1434168_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 437463 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 100580537 - -
          Illumina mouse-6 v1.0 expression beadchip 6550546 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006550546 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1243077 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243077 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620089 - -
          mouse-6 v1.1 (Illumina) scl53091.6_76-S - -
          mouse-6 v1.1 (Illumina) scl00226153.2_99-S - -
          Illumina Sentrix mouse-6.v1 bead array 6550546 - -
          Illumina Sentrix mouse-6.v1 bead array 100580537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00226153.2_99-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550546 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100580537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1243077 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243077 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913845 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620089 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline