Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468213_m1
          See other CELA2A GE Assays ›
          Gene Symbol:
          Cela2a
          Gene Name:
          chymotrypsin-like elastase family, member 2A
          Gene Aliases:
          Ela-2, Ela2, Ela2a
          Chromosome Location:
          Chr.4: 141814963 - 141826003 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cela2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007919.2 NP_031945.1 5 - 6 520 74
          GenBank mRNA BC026552.1 - 5 - 6 509 4990174 74
          X04573.1 - 5 - 6 520 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004226.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cela2a

          Panther Classification:

          Molecular Function -

          serine protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          establishment of skin barrier
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          serine hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 NM_007919.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 BC026552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 X04573.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 NM_007919.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 BC026552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P504538 X04573.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430799 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 638392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833113 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 716348 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151483 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144997 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154351 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 NM_007919.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 BC026552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 X04573.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 NM_007919.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 BC026552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P504538 X04573.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 NM_007919.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 BC026552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 X04573.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 NM_007919.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 BC026552.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504538 X04573.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23366181 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01008124 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144997 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154351 - -
          Whole Mouse Genome G4122A A_51_P504538 NM_007919.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504538 BC026552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504538 X04573.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504538 NM_007919.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504538 BC026552.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504538 X04573.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130843 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129946 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154351 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144997 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03706735 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03342377 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03342374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23366124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03677752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03706735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03677747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03891923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03343565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03435720 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03343561 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366156 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03772238 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924523 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03342376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03638510 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04086142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03363003 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03343564 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03935388 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03342374 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03924524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23366188 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518056 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518058 - -
          Affymetrix HT MG-430 PM Array Plate 1448281_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518050 - -
          Mouse Genome 430 2.0 Array 1448281_PM_a_at - -
          Mouse Genome 430 2.0 Array 1448281_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13706_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13706_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400032325.mm.1 TC0400004226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032325.mm.1 NM_007919.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035214.mm.1 TC0400004226.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035214.mm.1 NM_007919.3 Refseq
          Mouse Expression Set 430 1448281_PM_a_at - -
          Mouse Expression Set 430 1448281_a_at - -
          Murine Genome U74v2 Set 94037_at - -
          Murine Genome U74v2 Set 94037_PM_at - -
          Murine Genome U74v2 Set 167024_PM_at - -
          Murine Genome U74v2 Set 167024_at - -
          Mouse Genome 430A 2.0 Array 1448281_a_at - -
          Mouse Genome 430A 2.0 Array 1448281_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448281_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1448281_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 912383 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5270129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005270129 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674620 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674620 - -
          mouse-6 v1.1 (Illumina) scl013706.2_82-S - -
          Illumina Sentrix mouse-6.v1 bead array 5270129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl013706.2_82-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5270129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674620 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674620 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline