Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468248_m1
          See other ENDOG GE Assays ›
          Gene Symbol:
          Endog
          Gene Name:
          endonuclease G
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 30171401 - 30174069 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468248_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Endog
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007931.1 NP_031957.1 2 - 3 655 104
          GenBank mRNA AB012108.1 - 2 - 3 655 104
          AK144459.1 - 2 - 3 687 104
          BC030177.1 - 2 - 3 664 5029633 104
          X99394.1 - 2 - 3 151 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000528.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Endog

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          DNA metabolic process
          DNA catabolic process
          apoptotic DNA fragmentation
          response to tumor necrosis factor
          positive regulation of apoptotic process
          response to antibiotic
          nucleic acid phosphodiester bond hydrolysis
          positive regulation of hydrogen peroxide-mediated programmed cell death
          positive regulation of apoptotic DNA fragmentation
          nucleic acid binding
          nuclease activity
          endonuclease activity
          deoxyribonuclease activity
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101725 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101730 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 BC030177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 X99394.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 NM_007931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 AB012108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 BC030177.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 X99394.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 NM_007931.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 AB012108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625077 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 15795 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 BC030177.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 X99394.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 NM_007931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 AB012108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 AK144459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 BC030177.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 X99394.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 NM_007931.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 AB012108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 BC030177.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 X99394.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 NM_007931.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 AB012108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 BC030177.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 X99394.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 NM_007931.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 AB012108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101734 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101737 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101744 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21101731 - -
          Whole Mouse Genome G4122A A_51_P349073 BC030177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 X99394.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 NM_007931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 AB012108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 AK144459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 BC030177.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 X99394.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 NM_007931.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 AB012108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P349073 AK144459.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125798 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01758753 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01758749 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101725 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101727 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101729 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21101730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21101774 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101718 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01758755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01334552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02026206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01628488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01456082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01352845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101742 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01758756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02104202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01740382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02165668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01334553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01352848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21101723 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470953 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470951 - -
          Affymetrix HT MG-430 PM Array Plate 1421097_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438317_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470950 - -
          Affymetrix Mouse Gene 1.1 ST Array 10481420 - -
          Mouse Genome 430 2.0 Array 1421097_PM_at - -
          Mouse Genome 430 2.0 Array 1421097_at - -
          Mouse Genome 430 2.0 Array 1438317_PM_a_at - -
          Mouse Genome 430 2.0 Array 1438317_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13804_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13804_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200004126.mm.1 NM_007931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200004126.mm.1 NM_007931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200004126.mm.1 TC0200000528.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200004127.mm.1 NM_007931.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200004127.mm.1 NM_007931.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200004127.mm.1 TC0200000528.mm.1 MTA TC
          Mouse Expression Set 430 1421097_PM_at - -
          Mouse Expression Set 430 1438317_a_at - -
          Mouse Expression Set 430 1438317_PM_a_at - -
          Mouse Expression Set 430 1421097_at - -
          Murine Genome U74v2 Set 160494_at - -
          Murine Genome U74v2 Set 160494_PM_at - -
          Mouse Genome 430A 2.0 Array 1438317_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1421097_PM_at - -
          Mouse Genome 430A 2.0 Array 1421097_at - -
          Mouse Genome 430A 2.0 Array 1438317_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421097_at - -
          Affymetrix HT Mouse Genome 430A Array 1438317_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1438317_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421097_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 436144 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610735 - -
          Illumina mouse-6 v1.0 expression beadchip 3610735 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610735 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1244281 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230557 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244281 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230557 - -
          mouse-6 v1.1 (Illumina) scl21095.3.1_3-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610735 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21095.3.1_3-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610735 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244281 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21095.3.1_3-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline