Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00468329_m1
          See other LCN5 GE Assays ›
          Gene Symbol:
          Lcn5
          Gene Name:
          lipocalin 5
          Gene Aliases:
          E-RABP, Erabp, MEP10
          Chromosome Location:
          Chr.2: 25657932 - 25662230 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00468329_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lcn5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001042630.2 NP_001036095.1 2 - 3 286 64
          NM_001276257.1 NP_001263186.1 2 - 3 286 64
          NM_007947.3 NP_031973.2 2 - 3 286 64
          GenBank mRNA AK021014.1 - 2 - 3 284 64
          AK136629.1 - 2 - 3 289 64
          AK162355.1 - 2 - 3 285 64
          U68546.1 - 2 - 3 182 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000428.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lcn5

          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          transport
          retinoic acid metabolic process
          transporter activity
          retinoid binding
          small molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963727 NM_007947.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963727 AK136629.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963727 AK162355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963727 U68546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1963727 NM_001042630.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 564555 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AF151691.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AK006905.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AK145244.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AK154537.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AY515316.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 AY515317.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 BC057575.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 BC082278.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 837665 NM_011972.1 refseq_mrna_mouse_NM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128366 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963727 NM_007947.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963727 AK136629.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963727 AK162355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963727 U68546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1963727 NM_001042630.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191875 NM_007947.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P191875 U68546.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21073647 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128366 - -
          Whole Mouse Genome G4122A A_51_P191875 NM_007947.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P191875 U68546.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112909 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121716 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128366 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486596 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486572 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486576 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01486578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21073662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01486567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01872577 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02186806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01486578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02186803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02186804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522250 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01486592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01486587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02186808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02128529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21073658 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10470155 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470157 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470156 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470154 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10470150 - -
          Affymetrix HT MG-430 PM Array Plate 1419336_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10470149 - -
          Mouse Genome 430 2.0 Array 1419336_PM_at - -
          Mouse Genome 430 2.0 Array 1419336_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13863_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13863_at - -
          Affymetrix Mouse Clariom S Array PSR0200002614.mm.1 NM_001042630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002614.mm.1 NM_001276257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002614.mm.1 NM_007947.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002614.mm.1 TC0200000428.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002613.mm.1 NM_001042630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002613.mm.1 NM_001276257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002613.mm.1 NM_007947.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002613.mm.1 TC0200000428.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002612.mm.1 NM_001042630.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002612.mm.1 NM_001276257.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002612.mm.1 NM_007947.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002612.mm.1 TC0200000428.mm.1 MTA TC
          Mouse Expression Set 430 1419336_at - -
          Mouse Expression Set 430 1419336_PM_at - -
          Murine Genome U74v2 Set 99236_PM_at - -
          Murine Genome U74v2 Set 99236_at - -
          Mouse Genome 430A 2.0 Array 1419336_at - -
          Mouse Genome 430A 2.0 Array 1419336_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419336_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419336_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 469777 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3940082 - -
          Illumina mouse-6 v1.0 expression beadchip 3940082 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940082 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2588852 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2588852 - -
          mouse-6 v1.1 (Illumina) scl21169.5.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940082 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21169.5.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940082 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2588852 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2588852 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline