Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469263_m1
          See other H2-M3 GE Assays ›
          Gene Symbol:
          H2-M3
          Gene Name:
          histocompatibility 2, M region locus 3
          Gene Aliases:
          H-2M3, Hmt, M3a, R4B2
          Chromosome Location:
          Chr.17: 37270234 - 37274485 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469263_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-M3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013819.2 NP_038847.1 5 - 6 1026 72
          GenBank mRNA BC031984.1 - 5 - 6 1017 4007538 72
          U18797.1 - 5 - 6 998 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000765.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-M3

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          alpha-beta T cell activation involved in immune response
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of endogenous peptide antigen via MHC class Ib
          antigen processing and presentation of exogenous peptide antigen via MHC class Ib
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          positive regulation of tolerance induction
          positive regulation of T cell tolerance induction
          immune response-inhibiting cell surface receptor signaling pathway
          positive regulation of interferon-gamma production
          positive regulation of interleukin-12 production
          negative regulation of T cell proliferation
          defense response to bacterium
          positive regulation of nitric oxide biosynthetic process
          positive regulation of regulatory T cell differentiation
          positive regulation of natural killer cell mediated cytotoxicity
          negative regulation of dendritic cell differentiation
          receptor binding
          peptide antigen binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 U18797.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 NM_013819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 AI451265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 U18797.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 NM_013819.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 AI451265.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 949020 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P113749 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 U18797.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 NM_013819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 AI451265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 BC031984.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 U18797.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 NM_013819.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 AI451265.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 U18797.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 NM_013819.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 AI451265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 U18797.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 NM_013819.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 AI451265.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168115 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168113 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168126 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31168121 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P113749 - -
          Whole Mouse Genome G4122A A_52_P490263 U18797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490263 NM_013819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490263 AB267096.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P490263 CA481305.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 U18797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 NM_013819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 AI451265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 BC031984.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 U18797.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 NM_013819.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 AI451265.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P469968 BC031984.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111061 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P113749 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13091089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31168157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13300876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168115 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13334493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168118 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13141146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13278007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13091088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13156474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168122 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168120 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168117 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31168144 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10445124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445126 - -
          Affymetrix Mouse Gene 1.0 ST Array 10445121 - -
          Affymetrix HT MG-430 PM Array Plate 1421358_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10445119 - -
          Mouse Genome 430 2.0 Array 1421358_at - -
          Mouse Genome 430 2.0 Array 1421358_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14991_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14991_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700007698.mm.1 NM_013819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007698.mm.1 NM_013819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007698.mm.1 TC1700000765.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007696.mm.1 NM_013819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007696.mm.1 NM_013819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007696.mm.1 TC1700000765.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700007697.mm.1 NM_013819.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007697.mm.1 NM_013819.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700007697.mm.1 TC1700000765.mm.1 MTA TC
          Mouse Expression Set 430 1421358_PM_at - -
          Mouse Expression Set 430 1421358_at - -
          Murine Genome U74v2 Set 102730_at - -
          Murine Genome U74v2 Set 102731_PM_g_at - -
          Murine Genome U74v2 Set 102730_PM_at - -
          Murine Genome U74v2 Set 102731_g_at - -
          Mouse Genome 430A 2.0 Array 1421358_at - -
          Mouse Genome 430A 2.0 Array 1421358_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1421358_at - -
          Affymetrix HT Mouse Genome 430A Array 1421358_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 807637 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6100309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006100309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2670508 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2670508 - -
          mouse-6 v1.1 (Illumina) scl50733.5.1_21-S - -
          Illumina Sentrix mouse-6.v1 bead array 6100309 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50733.5.1_21-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6100309 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2670508 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2670508 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline