Search
Search

Exoc7| Gene Symbol: Exoc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162872.1 | NP_001156344.1 | 5 - 6 | 701 | 62 | |
| NM_016857.2 | NP_058553.2 | 5 - 6 | 701 | 62 | ||
| GenBank mRNA | AF014461.1 | - | 5 - 6 | 636 | 62 | |
| AK087601.1 | - | 5 - 6 | 689 | 62 | ||
| AK140450.1 | - | 5 - 6 | 675 | 62 | ||
| AK162868.1 | - | 5 - 6 | 687 | 62 | ||
| AK167769.1 | - | 5 - 6 | 684 | 62 | ||
| AK169928.1 | - | 5 - 6 | 688 | 62 | ||
| AK173094.1 | - | 5 - 6 | 677 | 62 | ||
| BC028927.1 | - | 5 - 6 | 677 | 4208803 | 62 | |
| BC053710.1 | - | 5 - 6 | 688 | 30056570 | 62 | |
Set Membership: |
|