Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469499_m1
          See other HEBP2 GE Assays ›
          Gene Symbol:
          Hebp2
          Gene Name:
          heme binding protein 2
          Gene Aliases:
          SOUL
          Chromosome Location:
          Chr.10: 18540123 - 18546076 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469499_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hebp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019487.3 NP_062360.1 3 - 4 700 89
          GenBank mRNA AF117614.1 - 3 - 4 630 89
          AK078041.1 - 680 89
          AK078130.1 - 3 - 4 700 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001807.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hebp2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of mitochondrial membrane potential
          positive regulation of necrotic cell death
          positive regulation of mitochondrial membrane permeability
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 AK078130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 BC006898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 AF117614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 NM_019487.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 AK078130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 BC006898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 AF117614.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P159612 NM_019487.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 332536 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759505 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409399 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 821988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 43828 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163669 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 AK078130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 BC006898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 AF117614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 NM_019487.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 AK078130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 BC006898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 AF117614.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P159612 NM_019487.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26976161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163669 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06990776 - -
          Whole Mouse Genome G4122A A_51_P159612 AK078130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 BC006898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 AF117614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 NM_019487.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 AK078130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 BC006898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 AF117614.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P159612 NM_019487.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112163 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08447009 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08207574 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08207577 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08207579 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08207571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26976149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08598200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08642875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08538983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08342599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08910528 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08538979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08752077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08307201 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26976181 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368099 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368100 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368098 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10368093 - -
          Affymetrix HT MG-430 PM Array Plate 1418917_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1446415_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1449271_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10368092 - -
          Mouse Genome 430 2.0 Array 1449271_a_at - -
          Mouse Genome 430 2.0 Array 1446415_at - -
          Mouse Genome 430 2.0 Array 1449271_PM_a_at - -
          Mouse Genome 430 2.0 Array 1418917_PM_at - -
          Mouse Genome 430 2.0 Array 1418917_at - -
          Mouse Genome 430 2.0 Array 1446415_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56016_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56016_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000013135.mm.1 NM_019487.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013135.mm.1 NM_019487.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013135.mm.1 TC1000001807.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000013136.mm.1 NM_019487.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013136.mm.1 NM_019487.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000013136.mm.1 TC1000001807.mm.1 MTA TC
          Mouse Expression Set 430 1449271_PM_a_at - -
          Mouse Expression Set 430 1418917_PM_at - -
          Mouse Expression Set 430 1449271_a_at - -
          Mouse Expression Set 430 1418917_at - -
          Mouse Expression Set 430 1446415_PM_at - -
          Mouse Expression Set 430 1446415_at - -
          Mouse Genome 430A 2.0 Array 1418917_PM_at - -
          Mouse Genome 430A 2.0 Array 1449271_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1418917_at - -
          Mouse Genome 430A 2.0 Array 1449271_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1418917_at - -
          Affymetrix HT Mouse Genome 430A Array 1418917_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449271_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1449271_a_at - -
          Affymetrix HT Mouse Genome 430B Array 1446415_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1446415_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 465508 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 105290075 - -
          MouseWG-6 v2 Expression BeadChip 1990309 - -
          Illumina mouse-6 v1.0 expression beadchip 105290075 - -
          Illumina mouse-6 v1.0 expression beadchip 1990309 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001990309 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2710455 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254381 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2710455 - -
          mouse-6 v1.1 (Illumina) scl38162.4_162-S - -
          Illumina Sentrix mouse-6.v1 bead array 1990309 - -
          Illumina Sentrix mouse-6.v1 bead array 105290075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38162.4_162-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1990309 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105290075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2710455 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254381 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2710455 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline