Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469712_m1
          See other CXCL16 GE Assays ›
          Gene Symbol:
          Cxcl16
          Gene Name:
          chemokine (C-X-C motif) ligand 16
          Gene Aliases:
          0910001K24Rik, AV290116, BB024863, CXCL16v1, CXCL16v2, SR-PSOX, Zmynd15, b2b498Clo
          Chromosome Location:
          Chr.11: 70454234 - 70459984 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469712_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxcl16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023158.6 NP_075647.3 3 - 4 835 67
          GenBank mRNA AF277001.1 - 3 - 4 298 67
          AF301017.1 - 3 - 4 364 67
          AK003085.1 - 3 - 4 494 67
          AK009599.1 - 3 - 4 384 67
          AK028875.1 - 3 - 4 837 67
          AK030515.1 - 3 - 4 260 67
          AK150224.1 - 3 - 4 523 67
          AK151330.1 - 3 - 4 609 67
          AK152102.1 - 3 - 4 609 67
          AK152547.1 - 3 - 4 787 67
          AK153192.1 - 3 - 4 551 67
          BC019961.1 - 3 - 4 716 3988542 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003097.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxcl16

          Panther Classification:

          Molecular Function -

          chemokine cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          receptor-mediated endocytosis
          chemotaxis
          T cell chemotaxis
          positive regulation of cell growth
          positive regulation of cell migration
          response to cytokine
          response to interferon-gamma
          response to tumor necrosis factor
          lymphocyte chemotaxis
          cell chemotaxis
          cellular response to lipopolysaccharide
          low-density lipoprotein receptor activity
          scavenger receptor activity
          cytokine activity
          chemokine activity
          chemokine receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900832 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900882 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P99888 AK028875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P99888 NM_023158.6 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P99888 AK028875.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P99888 NM_023158.6 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 634577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 186716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 931878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 50910 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105809 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P99888 AK028875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P99888 NM_023158.6 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P99888 AK028875.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P99888 NM_023158.6 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374203 BC019961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374203 GQ380017.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374203 AK152102.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374203 AK152547.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P374203 AK153192.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P99888 AK028875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P99888 NM_023158.6 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P99888 AK028875.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P99888 NM_023158.6 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900837 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27900846 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105809 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07388585 - -
          Whole Mouse Genome G4122A A_52_P99888 AK028875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P99888 NM_023158.6 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P99888 AK028875.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P99888 NM_023158.6 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374203 BC019961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374203 GQ380017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374203 AK152102.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374203 AK152547.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P374203 AK153192.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114820 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105809 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418250 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418238 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418251 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418245 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418248 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09418254 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900832 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900833 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27900881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27900753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900883 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900877 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09112640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09418254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09418238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09596430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09483950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27900858 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10387891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387895 - -
          Affymetrix HT MG-430 PM Array Plate 1449195_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1418718_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10387890 - -
          Affymetrix Mouse Gene 1.1 ST Array 10377826 - -
          Mouse Genome 430 2.0 Array 1418718_PM_at - -
          Mouse Genome 430 2.0 Array 1418718_at - -
          Mouse Genome 430 2.0 Array 1449195_PM_s_at - -
          Mouse Genome 430 2.0 Array 1449195_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66102_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66102_at - -
          Affymetrix Mouse Clariom S Array PSR1100028611.mm.1 NM_023158.6.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028611.mm.1 NM_023158.6 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028611.mm.1 TC1100003097.mm.1 MTA TC
          Mouse Expression Set 430 1418718_at - -
          Mouse Expression Set 430 1418718_PM_at - -
          Mouse Expression Set 430 1449195_s_at - -
          Mouse Expression Set 430 1449195_PM_s_at - -
          Murine Genome U74v2 Set 134771_at - -
          Murine Genome U74v2 Set 134771_PM_at - -
          Mouse Genome 430A 2.0 Array 1418718_at - -
          Mouse Genome 430A 2.0 Array 1449195_s_at - -
          Mouse Genome 430A 2.0 Array 1449195_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1418718_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418718_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418718_at - -
          Affymetrix HT Mouse Genome 430A Array 1449195_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1449195_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 713924 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 510278 - -
          MouseWG-6 v2 Expression BeadChip 101780301 - -
          Illumina mouse-6 v1.0 expression beadchip 101780301 - -
          Illumina mouse-6 v1.0 expression beadchip 510278 - -
          Illumina mouse-6 v1.0 expression beadchip 100450458 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2687586 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254723 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2687586 - -
          mouse-6 v1.1 (Illumina) scl0066102.2_212-S - -
          mouse-6 v1.1 (Illumina) scl41338.15.1_31-S - -
          mouse-6 v1.1 (Illumina) scl39995.5_256-S - -
          Illumina Sentrix mouse-6.v1 bead array 101780301 - -
          Illumina Sentrix mouse-6.v1 bead array 100450458 - -
          Illumina Sentrix mouse-6.v1 bead array 510278 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0066102.2_212-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101780301 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100450458 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2687586 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254723 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2461299 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2687586 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0066102.2_212-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline