Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00469759_m1
          See other RTP4 GE Assays ›
          Gene Symbol:
          Rtp4
          Gene Name:
          receptor transporter protein 4
          Gene Aliases:
          5830458K16Rik, Ifrg28
          Chromosome Location:
          Chr.16: 23601379 - 23614222 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00469759_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rtp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023386.5 NP_075875.3 1 - 2 232 59
          GenBank mRNA AJ251364.1 - 1 - 2 227 59
          AK007477.1 - 1 - 2 210 59
          AK018021.1 - 1 - 2 225 59
          AY562228.1 - 1 - 2 162 59
          BC024872.1 - 1 - 2 190 3990099 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000346.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rtp4

          Gene Ontology Categories:

          Function(s) Process(es)

          detection of chemical stimulus involved in sensory perception of bitter taste
          protein targeting to membrane
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 BC024872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 BC006590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 AJ251364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 AK007477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 NM_023386.5 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 BC024872.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 BC006590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 AJ251364.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 AK007477.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304170 NM_023386.5 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966882 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 490456 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P175674 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 BC024872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 BC006590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 AJ251364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 AK007477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 NM_023386.5 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 BC024872.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 BC006590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 AJ251364.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 AK007477.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304170 NM_023386.5 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30603967 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P175674 - -
          Whole Mouse Genome G4122A A_51_P304170 BC024872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 BC006590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 AJ251364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 AK007477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 NM_023386.5 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 BC024872.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 BC006590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 AJ251364.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 AK007477.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304170 NM_023386.5 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130534 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138226 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P175674 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12905295 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12905298 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12905300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12905299 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12814392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30603910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30604008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12689755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12695226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12512525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12582792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12905300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12833292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603987 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12905297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12924284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604000 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12512529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12878137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12878129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12814391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12512517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12695222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604004 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12905302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12950739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12496485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30604022 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12905301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12551369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30603997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10434780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434781 - -
          Affymetrix Mouse Gene 1.0 ST Array 10434779 - -
          Affymetrix HT MG-430 PM Array Plate 1449143_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418580_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10434778 - -
          Mouse Genome 430 2.0 Array 1418580_PM_at - -
          Mouse Genome 430 2.0 Array 1449143_PM_at - -
          Mouse Genome 430 2.0 Array 1418580_at - -
          Mouse Genome 430 2.0 Array 1449143_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67775_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67775_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600003431.mm.1 NM_023386.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003431.mm.1 NM_023386.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003431.mm.1 TC1600000346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003433.mm.1 NM_023386.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003433.mm.1 NM_023386.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003433.mm.1 TC1600000346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600003432.mm.1 NM_023386.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003432.mm.1 NM_023386.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1600003432.mm.1 TC1600000346.mm.1 MTA TC
          Mouse Expression Set 430 1418580_PM_at - -
          Mouse Expression Set 430 1418580_at - -
          Mouse Expression Set 430 1449143_PM_at - -
          Mouse Expression Set 430 1449143_at - -
          Mouse Genome 430A 2.0 Array 1418580_at - -
          Mouse Genome 430A 2.0 Array 1418580_PM_at - -
          Mouse Genome 430A 2.0 Array 1449143_at - -
          Mouse Genome 430A 2.0 Array 1449143_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449143_at - -
          Affymetrix HT Mouse Genome 430A Array 1418580_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449143_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418580_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 679589 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2060112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002060112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2776509 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2776509 - -
          mouse-6 v1.1 (Illumina) scl49298.8_323-S - -
          Illumina Sentrix mouse-6.v1 bead array 2060112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49298.8_323-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2060112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2873988 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2776508 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2873988 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0